Skip to Content
Merck
All Photos(1)

Documents

MABT1503

Sigma-Aldrich

Anti-PGM5 (Aciculin) Antibody, clone 14F8

clone 14F8, from mouse

Synonym(s):

Aciculin, Phosphoglucomutase-like Protein 5, PGM-RP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

14F8, monoclonal

species reactivity

human, rabbit, rat, bovine, mouse

species reactivity (predicted by homology)

frog, avian, porcine

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PGM5(5239)

General description

Phosphoglucomutase-like protein 5 (UniProt: Q15124; also known as Aciculin, Phosphoglucomutase-related protein, PGM-RP) is encoded by the PGM5 (also known as PGMRP) gene (Gene ID: 5239) in human. Aciculin is a dystrophin and utrophin-binding protein that is highly homologous to the glycolytic enzyme phosphoglucomutase type 1. However, it lacks phosphoglucomutase activity. Aciculin is a component of adherens-type cell-cell and cell-matrix junctions and is concentrated in focal contacts at the ends of actin bundles and associates with actin filaments. Aciculin is also reported to be an interaction partner of Filamin C and Xin proteins. Higher levels of Aciculin are reported in smooth and cardiac muscle and lower levels are present in skeletal muscle, liver, kidney and brain tissue. In striated muscle, it mainly localizes to the intercalated discs of the heart and to myotendinous junctions. Low levels of Aciculin are expressed in 10-12-week-old fetus and these levels increase progressively into infancy reaching maximum levels at maturity. Two isoforms of Aciculin have been reported that are produced by alternative splicing. Aciculin knockdown in myotubes is shown to cause failure in myofibril assembly, alignment and membrane attachment, and a massive reduction in myofibril number. (Ref.: Molt, S., et al., (2014). J. Cell Sci.127(16); 3578-3592).

Specificity

Clone 14F8 detects Phosphoglucomutase-like protein 5 in mouse and rat cardiac tissue and is predicted to react with human.

Immunogen

Cytoskeletal fraction of the human myometrium tissue extract.

Application

Anti-PGM5 (Aciculin), clone 14F8, Cat. No. MABT1503, is a mouse monoclonal antibody that detects Phosphoglucomutase-like protein 5 and has been tested for use in ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: 1 µg/mL from a representative lot detected PGM5 (Aciculin) in rat heart tissue lysate.

Immunoprecipitation Analysis: A representative lot immunoprecipitated PGM5 (Aciculin) in Immunoprecipitation applications (Belkin, A.M., et. al. (1995). J Biol Chem. 270(11):6328-37; Belkin, A.M., et. al. (1995). Exp Cell Res. 221(1):132-40).

Immunofluorescence Analysis: A representative lot detected PGM5 (Aciculin) in Immunofluorescence applications (Belkin, A.M., et. al. (1994). J Cell Sci. 107 ( Pt 7):1993-2003).

Immunohistochemistry Analysis: A representative lot detected PGM5 (Aciculin) in Immunohistochemistry applications (Belkin, A.M., et. al. (1995). Exp Cell Res. 221(1):132-40).

Western Blotting Analysis: A representative lot detected PGM5 (Aciculin) in Western Blotting applications (Belkin, A.M., et. al. (1995). J Biol Chem. 270(11):6328-37; Belkin, A.M., et. al. (1994). J Cell Sci. 107 ( Pt 7):1993-2003).

ELISA Analysis: A representative lot detected PGM5 (Aciculin) in ELISA applications (Molt, S., et. al. (2014). J Cell Sci. 127(Pt 16):3578-92).

Quality

Evaluated by Western Blotting in mouse heart tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PGM5 (Aciculin) in mouse heart tissue lysate.

Target description

~57 kDa observed; 62.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service