Skip to Content
Merck

Skip To

MABS519

Anti-P40 Antibody, clone 11F12.1

clone 11F12.1, from mouse

Synonym(s):

Tumor protein 63, p63, Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, Transformation-related protein 63, TP63, Tumor protein p73-like, p73L, p40, p51

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 18, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,900.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
11F12.1, monoclonal
Application:
IHC, WB
Citations:
-

CZK 10,900.00


Estimated to ship onJune 18, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11F12.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TP63(8626)

General description

P40, also known as Tumor protein 63, p63, Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor , KET, Transformation-related protein 63, TP63, Tumor protein p73-like, p73L, p40, or p51, and encoded by the gene TP63/KET/P63/P73H/P73L/TP73L, is a transcriptional activator or repressor depending upon the target gene and the isoform involved. P40 has multiple isoforms. P40 is involved in Notch signaling and plays a role in p53/TP53 dependent apoptosis and epithelial morphogenesis. Moreover P40, developmentally, is required for proper limb formation. P40 is widely expressed in the adult and during development. Some isoforms, such as isoform 10 are primarily expressed in cancerous tissues while others are found only in normal tissues. Mutations in P40 lead to a variety of dysplasias and malformations, including Acro-dermato-ungual-lacrimal-tooth syndrome and Ankyloblepharon-ectodermal defects cleft lip/palate disorder and several more tragic disfiguring syndromes.
~40 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human P40.

Application

Anti-P40 Antibody, clone 11F12.1 is an antibody against P40 for use in IHC (Paraffin) & western blotting.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected P40 in human prostate and human kidney tissue.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in SK-OV-3 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected P40 in 10 µg of SK-OV-3 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB56000289AMAB91224AB9985
clone

11F12.1, monoclonal

clone

RM383, monoclonal

clone

CL3748, monoclonal

clone

polyclonal

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

biological source

mouse

biological source

-

biological source

mouse

biological source

rabbit

antibody form

purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

Gene Information

human ... TP63(8626)

Gene Information

human ... TP63(8626)

Gene Information

human ... TP63(8626)

Gene Information

human ... TP53(7157)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters