MABS449
Anti-Acylation Stimulating Protein Antibody, clone 4H3
clone 4H3, from mouse
Synonym(s):
Complement C3, C3adesArg, C3, PZP-like alpha-2-macroglobulin domain-containing protein 1
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
4H3, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... C3(718)
General description
Immunogen
Application
Signaling
Metabolism
Insulin/Energy Signaling
Lipid Metabolism & Weight Regulation
Quality
Western Blotting Analysis: 0.1 µg/mL of this antibody detected purified Acylation Stimulating Protein from transfected HEK293 cell.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service