Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship onJune 10, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 10,500.00 |
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4D2.2, monoclonal
Application:
IHC, WB
Citations:
-
conjugate
unconjugated
Quality Segment
antibody form
purified antibody
clone
4D2.2, monoclonal
purified by
using protein G
species reactivity
mouse, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgG1κ
UniProt accession no.
target post-translational modification
unmodified
General description
GST-tagged recombinant protein corresponding to an internal fragment from the C-terminal half of human CLCA1.
~83 kDa observed. 125 kDa glycosylated precursor form, 90 kDa and 37-41 kDa processed/cleaved fragements reported (Gruber, A.D., et al. (1998). Genomics. 54(2):200-214).
Application
Epitope: Internal (C-terminal half).
Biochem/physiol Actions
Detects the 90 kDa processed/cleaved 4-transmembrane glycoprotein, but not the cleaved 37-41 kDa C-terminal fragment.
Physical form
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in human small intestine tissue lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CLCA1 in 10 µg of human small intestine tissue lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CLCA1 in 10 µg of human small intestine tissue lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.