Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Estimated to ship onJune 22, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 10,900.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
1G6, monoclonal
species reactivity
mouse, yeast, human
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MTM1(4534)
General description
Immunogen
Application
Signaling
Signaling Neuroscience
Western Blotting Analysis: A representative lot from an independent laboratory detected Myotubularin in lymphoblast lysates (Laporte, J., et al. (2002). J Cell Sci. 115(Pt 15):3105-3117.).
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Myotubularin from C2C12 cell lysates. (Laporte, J., et al. (2002). J Cell Sci. 115(Pt 15):3105-3117.).
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Myotubularin from C2C12 cells. (Buj-Bello, A. et al. (2002). Hum Mol Genet. 11(19):2297-2307.).
Physical form
Preparation Note
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Myotubularin in 10 µg of HeLa cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form ascites fluid | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form ascites fluid |
| biological source mouse | biological source rabbit | biological source rabbit | biological source mouse |
| species reactivity mouse, yeast, human | species reactivity human, mouse | species reactivity human | species reactivity human |
| clone 1G6, monoclonal | clone polyclonal | clone polyclonal | clone 1E11, monoclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Gene Information human ... MTM1(4534) | Gene Information human ... MTM1(4534) | Gene Information human ... MTM1(4534) | Gene Information human ... MTMR3(8897) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



