Skip to Content
Merck

Skip To

MABS341

Anti-LRP6 Antibody, clone 1C10

clone 1C10, from mouse

Synonym(s):

Low-density lipoprotein receptor-related protein 6, LRP-6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
CZK 10,900.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1C10, monoclonal
Application:
IP, WB
Citations:
-

CZK 10,900.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1952

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C10, monoclonal

species reactivity

human, rat, mouse

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LRP6(4040)

General description

Low density lipoprotein receptor-related protein 6, also known as LRP6, is a member of the low density Lipoprotein receptor gene family (LDLR). LDLR are cell surface proteins. LRP6 plays a crucial role in embryonic development by functioning as a co-receptor for the Wnt signaling pathway. LRP6 is highly homologous and presents itself as a dimer in the cell membrane until it is phosphlorylated by Wnt. LRP6 has a high affinity with the Dickkopf (DKK) family of antagonists which, when bound, block Wnt. When LRP6 binds to Wnt it is phosphlorated at multiple sites, which results in Axin, a scaffolding protein, being recruited to the cell membrane and β-catenin activation. β-catenin prevents Wnt and LRP6 from binding. LRP6 is associated with autosomal dominant coronary artery disease type 2. Mutations resulting in the omission of the extracellular domain of LPR6 results in constant activity whereas carboxy intracellular domain exclusion has a negative effect, resulting in the blocking of Wnt signaling.
~200 kDa observed

Immunogen

Flag-tagged recombinant protein corresponding to human LRP6.

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated LRP6 in certain cell lyates (Khan, Z., et al. (2007). Mol Cell Biol. 27(20):7291-7301.).
This LRP6 antibody is validated for use in WB & IP for the detection of the LRP6 protein.

Physical form

Format: Purified
Purified mouse monoclonal IgG containing PBS with 0.05% sodium azide.

Analysis Note

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LRP6 in 10 µg of HEK293 cell lysate. For enhanced results, it is recommended for end-users to perform higher dilutions of this antibody.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1300743MABS2001MWH0004041M1
Gene Information

human ... LRP6(4040)

Gene Information

human ... LRP6(4040)

Gene Information

human ... LRP1(4035)

Gene Information

human ... LRP5(4041)

species reactivity

human, rat, mouse

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human

clone

1C10, monoclonal

clone

polyclonal

clone

11H4, monoclonal

clone

1E9, monoclonal

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody form

purified antibody

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters