Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 μg | Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 3,880.00 |
100 μg | Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 9,780.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
KT10, monoclonal
mol wt
calculated mol wt 1.042 kDa
purified by
using Protein A
species reactivity
rat, mouse, human
species reactivity (predicted by homology)
bovine
packaging
antibody small pack of 100 μg
technique(s)
ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, inhibition assay: suitable
isotype
IgG2a
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
mouse ... Stag1(20842)
General description
Immunogen
Application
Evaluated by Immunocytochemistry in K562 cells.
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected PI(4,5)P2 (PIP2) in K562 cells.
Tested Applications
ELISA Analysis: A representative lot detected PIP2 in ELISA applications (Fukami, K. et al. (1988). Proc Natl Acad Sci USA. 85(23): 9057-61).
Immunocytochemistry Analysis: A representative lot detected PIP2 in Immunocytochemistry applications (Yoneda, A., et al. (2020). Biochem Biophys Res Commun. 527(4): 1050-1056).
Flow Cytometry Analysis: Analysis: A representative lot detected PIP2 in Flow Cytometry applications (Yoneda, A., et al. (2020). Biochem Biophys Res Commun. 527(4): 1050-1056).
Inhibition Assay: A representative lot of this antibody inhibited intracellular breakdown of PIP2 and also blocked the proliferation of Src- and erbB-transformed cells. (Fukami, K. et al. (1988). Proc Natl Acad Sci USA. 85(23): 9057-61).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form purified antibody | antibody form purified antibody | antibody form purified antibody | antibody form purified antibody |
| biological source mouse | biological source mouse | biological source rat | biological source mouse |
| species reactivity rat, mouse, human | species reactivity human | species reactivity mouse, human | species reactivity chicken, human, mouse |
| clone KT10, monoclonal | clone MAB243 (also referenced as L2), monoclonal | clone 7E7, monoclonal | clone 12C11, monoclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate - |
| mol wt calculated mol wt 1.042 kDa | mol wt calculated mol wt 53.01 kDa | mol wt calculated mol wt 41.91 kDa | mol wt calculated mol wt 170 kDa, observed mol wt ~170 kDa |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



