Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
C1096, monoclonal
Application:
ICC, IF, IHC, IP, WB
Citations:
-
conjugate
unconjugated
antibody form
purified antibody
clone
C1096, monoclonal
purified by
using protein G
species reactivity
mouse, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
UniProt accession no.
target post-translational modification
unmodified
General description
82.52 kDa calculated.
Sca-1+ treated Human cardiac progenitor cells (CPCs).
Application
Imunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected PECAM-1 (CD31) in human kidney tissue sections.
Flow Cytometry Analysis: 1 µg from a representative lot detected PECAM-1 (CD31) in Jurkat cells.
Immunofluorescence Analysis: A representative lot detected PECAM-1 (CD31) in Immunofluorescence applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunoprecipitation Analysis: A representative lot immunoprecipitated PECAM-1 (CD31) in Immunoprecipitation applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunocytochemistry Analysis: A representative lot detected PECAM-1 (CD31) in Immunocytochemistry applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Western Blotting Analysis: A representative lot detected PECAM-1 (CD31) in Western Blotting applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunohistochemistry (Paraffin) Analysis: A representative lot detected PECAM-1 (CD31) in Immunohistochemistry applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Flow Cytometry Analysis: 1 µg from a representative lot detected PECAM-1 (CD31) in Jurkat cells.
Immunofluorescence Analysis: A representative lot detected PECAM-1 (CD31) in Immunofluorescence applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunoprecipitation Analysis: A representative lot immunoprecipitated PECAM-1 (CD31) in Immunoprecipitation applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunocytochemistry Analysis: A representative lot detected PECAM-1 (CD31) in Immunocytochemistry applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Western Blotting Analysis: A representative lot detected PECAM-1 (CD31) in Western Blotting applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Immunohistochemistry (Paraffin) Analysis: A representative lot detected PECAM-1 (CD31) in Immunohistochemistry applications (Moerkamp, A.T., et. al. (2017). Stem Cells Dev. 26(21):1552-1565).
Biochem/physiol Actions
Clone C1096 is a mouse monoclonal antibody that specifically detects human PECAM-1.
Physical form
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected PECAM-1 (CD31) in human tonsil tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected PECAM-1 (CD31) in human tonsil tissue sections.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.