Skip to Content
Merck

MABS1933

Anti-METTL3 Antibody, clone 29C8

Synonym(s):

EC:2.1.1.348, MT-A70, Methyltransferase-like protein 3, N6-adenosine-methyltransferase 70 kDa subunit, N6-adenosine-methyltransferase catalytic subunit, hMETTL3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
29C8, monoclonal
Application:
ELISA, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

29C8, monoclonal

mol wt

calculated mol wt 64.47 kDa, observed mol wt ~65 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

bovine, porcine, canine, monkey, mouse

packaging

antibody small pack of 100

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2cκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... METTL3(56339)

General description

N6-adenosine-methyltransferase catalytic subunit (UniProt: Q86U44; also known as EC:2.1.1.348, Methyltransferase-like protein 3, hMETTL3, N6-adenosine-methyltransferase 70 kDa subunit, MT-A70) is encoded by the METTL3 (also known as MTA70) gene (Gene ID: 56339) in human. METTL3 heterodimerizes with METTL14 to form an antiparallel heterodimer, which constitutes an active methyltransferase that methylates adenosine residues at the N6 position of some RNAs and regulates various cellular processes, including differentiation of embryonic and hematopoietic stem cells, cortical neurogenesis, differentiation of T-cells, and primary miRNA processing. In response to UV caused DNA damage, METTL3 rapidly catalyzes the formation of m6A on poly(A) transcripts at DNA damage sites that leads to the recruitment of POLK to these sites. It is localized both in the cytoplasm and in the nucleus. It colocalizes with speckles in interphase nuclei. In response to UV irradiation, it is shown to colocalize to DNA damage sites. Its methyltransferase activity is inhibited by sumoylation. However, sumoylation does not affect its subcellular location or interaction with METTL14. Overexpression of METTL3 is reported in several human cancers and its knockdown is known to promote apoptosis. METTL3 is reported to be strongly and directly phosphorylated by ERK2 at serine 43, 50, and 525 and to a lesser extent by p38 and JNK. METTL3 phosphorylation increases its USP5-mediated deubiquitination that leads to its stabilization. Phosphorylation of METTL3 is reported to promote murine embryonic stem cell differentiation. Clone 29C8 is suitable for complex purification of both METTL3 and-14 and is useful in simultaneous analysis of both proteins. (Ref.: Sun, H-L., et al. (2020). Mol. Cell. 80(4); 633-647; Schöller, E., et al. (2018). RNA 24(4); 499-512; Wang, P., et al. (2016). Mol. Cell. 63(2); 306-317).

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 15 amino acid from the N-terminal region of human METTL3.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from wild-type C643 cells.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected METTL3 in lysate from wild-type C643 cells, but not in lysate from METTL3 knockout C643 cells.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated METTL3 in Immunoprecipitation applications (Scholler, E., et al. (2018). RNA. 24(4):499-512).

Western Blotting Analysis: A representative lot detected METTL3 in Western Blotting applications (Scholler, E., et al. (2018). RNA. 24(4):499-512).

ELISA Analysis: A representative lot detected METTL3 in ELISA applications (Scholler, E., et al. (2018). RNA. 24(4):499-512).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 29C8 is a mouse monoclonal antibody that detects METTL3. It targets an epitope within 15 amino acids from the N-terminal region.

Physical form

Purified mouse monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library