Skip to Content
Merck
All Photos(2)

Documents

MABS1190

Sigma-Aldrich

Anti-Glucokinase/GCK Antibody, clone 9G1.2

clone 9G1.2, from mouse

Synonym(s):

Glucokinase, GK, Hexokinase-4, Hexokinase-D, Hexokinase type IV, HK IV, HK4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9G1.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GCK(2645)

General description

Glucokinase (EC 2.7.1.2; UniProt P35557; also known as GK, Hexokinase-4, Hexokinase-D, Hexokinase type IV, HK IV, HK4) is encoded by the GCK (also known as HHF3, MODY2) gene (Gene ID 2645) in human. Glucokinase/GK functions as the body′s primary glucose sensor by catalyzing the ATP-dependent phosphorylation of glucose to glucose-6-phosphate, the rate-limiting step of glucose metabolism in human pancreas and liver. GK activity is regulated rapidly in response to the elevated blood glucose after feeding, whereby a change in GK conformation causes a dissociation of the nuclear GK-glucokinase regulatory protein (GKRP) complex, allowing GK translocation to the cytoplasm in an activated state. In mammals, there exist four types of hexokinases, HK1, HK2, and HK3 have high affinity for glucose, while GK (also known as HK4) has low affinity for glucose and its expression is restricted to the pancreas and liver. GCK gene mutations cause Maturity-onset diabetes of the young 2 (MODY2) and Familial hyperinsulinemic hypoglycemia 3 (HHF3).

Specificity

Clone 9G1.2 recognizes an epitope present in all three spliced isoforms of human Glucokinase/GCK reported by UniProt (P35557).

Immunogen

Epitope: Internal (C-terminal half).
GST-tagged recombinant internal fragment from the C-terminal half of human Glucokinase/GCK.

Application

Anti-GlucokinaseAntibody, clone 9G1.2 is an antibody against GCK for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Glucokinase/GCK in human liver, colon, and tonsil tissue sections.
Research Category
Signaling
Research Sub Category
Glucose/Glycogen Metabolism

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Glucokinase/GCK in 10 µg of HepG2 cell lysate.

Target description

~52 kDa observed. ~52 kDa observed. 52.19 kDa (isoform 1), 52.13 kDa (isoform 2), and 52.04 kDa (isoform 3) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service