Skip to Content
Merck
All Photos(1)

Key Documents

MABN851

Sigma-Aldrich

Anti-CRALBP Antibody, clone 3H9.1

clone 3H9.1, from mouse

Synonym(s):

Retinaldehyde-binding protein 1, Cellular retinaldehyde-binding protein, CRALBP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,300.00

CZK 10,300.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,300.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

CZK 10,300.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H9.1, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RLBP1(6017)

General description

Retinaldehyde-binding protein 1 (UniProt P12271; also known as Cellular retinaldehyde-binding protein) is encoded by the RLBP1 (also known as CRALBP) gene (Gene ID 6017) in human. CRALBP is a retinoid-binding protein expressed in retinal pigment epithelial (RPE) cells and Müller glia involved in the retinal visual cycle. Photoactivation of a visual pigment molecule in vertebrate rod and cone photoreceptors renders the pigment molecule into a bleached all-trans retinal form, which must be exported out of the photoreceptors to be converted back to the light-sensitive 11-cis retinal form in RPE cells (for both rods and cones) or in retinal Müller glia (for cones only). The 11-cis chromophore is then imported back into photoreceptors, where it combines with a free opsin to regenerate the visual pigment. CRALBP is a 36 kDa water-soluble protein that facilitates the intracellular transport of hydrophobic 11-cis retinoids. Mutations in human RLBP1 cause several autosomal recessive retinal diseases, such as retinitis pigmentosa, Bothnia dystrophy, retinitis punctata albescens, fundus albipunctatus, and Newfoundland rod-cone dystrophy. Likewise, Rlbp1-knockout mice exhibit M-opsin mislocalization, M-cone loss, and impaired cone-driven visual behavior and light responses. Viral expression of CRALBP in Müller cells, but not RPE cells, is reported to rescue the retinal visual cycle and M-cone sensitivity in Rlbp1-knockout mice.

Immunogen

Epitope: N-terminal half.
GST-tagged recombinant protein corresponding to the N-terminal half of human CRALBP.

Application

Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Quality

Evaluated by Western Blotting in mouse retina tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CRALBP in 10 µg of mouse retina tissue lysate.

Target description

~36 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service