Skip to Content
Merck
All Photos(5)

Key Documents

MABN833

Sigma-Aldrich

Anti-phospho Drebrin (Ser142), clone 3C14 Antibody

Anti-phospho Drebrin (Ser142), clone 3C14 Antibody
1 of 1 reviewers received a sample product or took part in a promotion

clone 3C14, from mouse

Synonym(s):

Drebrin, Developmentally-regulated brain protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,100.00

CZK 12,100.00


Estimated to ship onMarch 12, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 12,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 12,100.00


Estimated to ship onMarch 12, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C14, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer142)

Gene Information

human ... DBN1(1627)

General description

Developmentally-regulated brain protein, also known as Drebrin, and encoded by the gene name DBN1, is a developmentally-regulated neuron-specific protein, expressed during outgrowth and maturation of dendrites, and localized within dendrites of the adult brain. Drebrin is required for actin polymerization at immunological synapses (IS) and for CXCR4 recruitment to IS. Recent studies have shown that Drebrins may play a role in cell migration, extension of neuronal processes and plasticity of dendrites. Drebrins are classified into two forms of the embryonic type (E1 and E2) and one form of the adult type (A). The time course of their appearance are different from each other. Their structures are closely related. Adult rat brain expresses only drebrin A while drebrin E1 or E2 is observed in immature animals. Drebrin E+A- signals have been observed in the cell bodies of migrating neuroblasts in the core region; however, only fibrous and punctate drebrin E+A- signals have been observed in postmigratory neuroblasts at the outer layers. Studies have demonstrated that the disappearance of Drebrin E+A- signals from the cell body coincides with the cessation of neuronal migration. The disappearance of Drebrin E from the cell body may be a molecular switch for the cessation of migration in newly generated neuroblast. Drebrin is expressed mainly in brain neurons.

Immunogen

KLH-conjugated linear peptide corresponding to rat phospho Drebrin (Ser142).

Application

Anti-phospho Drebrin (Ser142), clone 3C14 | MABN833 is an antibody against phospho Drebrin for use in Western Blotting, Immunocytochemistry, Peptide Inhibition Assay.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected phospho Drebrin (Ser142) in 10 µg of rat neonatal brain tissue lysate.
Immunocytochemistry Analysis: 20 µg/mL from a representative lot detected phospho Drebrin (Ser142) in HeLa and NIH/3T3 cells.
Peptide Inhibition Analysis: 0.5 µg/mL from a representative lot detected phospho Drebrin (Ser142) in 10 µg of rat neonatal brain tissue lysate.
Immunocytochemistry Analysis: A representative lot detected phospho Drebrin (Ser142) in rat embryonic cortical culture (Prof. Phillip R. Gordon-Weeks, King′s College London).

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected phospho Drebrin (Ser142) in 10 µg of SH-SY5Y cell lysate.

Target description

~100 kDa observed. The calculated molecular weight is 71 kDa, however phospho Drebrin (Ser142) has been shown as a ~100 kDa band in western blots (Worth, D.C., et al. (2013). JCB. 202(5):793-806).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG3κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paloma Alvarez-Suarez et al.
International journal of molecular sciences, 22(17) (2021-09-11)
Proper muscle function depends on the neuromuscular junctions (NMJs), which mature postnatally to complex "pretzel-like" structures, allowing for effective synaptic transmission. Postsynaptic acetylcholine receptors (AChRs) at NMJs are anchored in the actin cytoskeleton and clustered by the scaffold protein rapsyn
Thanushiyan Poobalasingam et al.
Journal of neurochemistry, 160(2), 185-202 (2021-09-04)
Co-ordinating the dynamic behaviour of actin filaments (F-actin) and microtubules in filopodia is an important underlying process in neuritogenesis, but the molecular pathways involved are ill-defined. The drebrin/end-binding protein 3 (EB3) pathway is a candidate pathway for linking F-actin to

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. UK
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Helpful suggestion

    It would help prospective buyers if you included a list of research papers where this antibody has been used. For example:
    Dart, A. E., Worth, D. C., Muir, G., Chandra, A., Morris, J. D., McKee, C., Verrill, C., Bryant, R. J. and Gordon-Weeks, P. R. (2017) The drebrin/EB3 pathway drives invasive activity in prostate cancer. Oncogene, 36, 4111-4123.
    Trivedi, N., Stabley, D. R., Cain, B. et al. (2017) Drebrin-mediated microtubule-actomyosin coupling steers cerebellar granule neuron nucleokinesis and migration pathway selection. Nature Comms, 8, 14484.
    Uddin, B., Partscht, P., Chen, N. P. et al. (2019) The human phosphatase CDC14A modulates primary cilium length by regulating centrosomal actin nucleation. EMBO reports, 20(1):e46544.
    Poobalasingham, T., Bianco, F., Oozeer, F. and Gordon-Weeks, P. R. (2022) The drebrin/EB3 pathway regulates cytoskeletal dynamics to drive neuritogenesis in embryonic cortical neurons. J. Neurochem., 160, 185-202.
    Chen, C., Chu, C. H., Chu, Y. et al. (2022) Neuronal paxillin and drebrin mediate BDNF-induced force transduction and growth cone turning in a soft-tissue-like environment. Cell Reports, 40, 111188.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service