Skip to Content
Merck
All Photos(3)

Key Documents

MABN812

Sigma-Aldrich

Anti-VPS4A Antibody, clone 14B1.1

clone 14B1.1, from mouse

Synonym(s):

Vacuolar protein sorting-associated protein 4A, Protein SKD2, VPS4-1, hVPS4, VPS4A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,100.00

CZK 10,100.00


Estimated to ship onFebruary 17, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

CZK 10,100.00


Estimated to ship onFebruary 17, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14B1.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VPS4A(27183)

General description

Vacuolar protein sorting-associated protein 4A (EC 3.6.4.6; UniProt Q9UN37; also known as hVPS4, Protein SKD2, SKD1-homolog, Vacuolar protein sorting factor 4A, Vacuolar sorting protein 4, VPS4A) is encoded by the VPS4A gene (also known as SKD1A, VPS4, VPS4-1) (Gene ID 27183) in human. Vps4 is a member of the ATPases associated with various cellular activities. The ESCRT (endosomal sorting complexes required for transport) machinery consists of five complexes (SCRT-0, ESCRT-I, ESCRT-II, ESCRT-III, and Vps4) that mediate multiple cellular membrane remodeling events, ranging from multivesicular bodies and exosomes formation to the final membrane abscission stage of cytokinesis, all of which converge in the scission of the narrow membrane neck that connects bud to parent membrane or daughter cell to daughter cell. Scission depends on the polymerization of soluble ESCRT-III monomers or dimers into tightly membrane-bound filaments. Vps4 is essential for ESCRT function in cells and it mediates the disassembly of ESCRT-III filaments to ensure a constant replenishing of the pool of soluble ESCRT-III monomers. The ESCRTs are also exploited by HIV-1 and many other enveloped viruses for their escape from the host cells. Vps4A is reported to facilitate the secretion of oncogenic miRNAs in exosomes and downregulation of Vps4A in hepatocellular carcinoma (HCC) is associated with tumor progression and metastasis.

Immunogen

Epitope: Near N-terminus
GST-tagged recombinant protein corresponding to human VPS4A sequence at the N-terminus.

Application

Detect VPS4A using this Anti-VPS4A Antibody, clone 14B1.1 validated for use in Western Blotting and Immunohistochemistry (Paraffin).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected VPS4A in 200 µg of MCF-7 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected VPS4A in rat cerebral cortex tissue.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected VPS4A in 200 µg of Jurkat cell lysate.

Target description

~50 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service