Skip to Content
Merck
All Photos(3)

Key Documents

MABN48

Sigma-Aldrich

Anti-UCHL1 Antibody, clone MCA-BH7

clone MCA-BH7, from mouse

Synonym(s):

Ubiquitin carboxyl-terminal hydrolase isozyme L1, UCH-L1, Neuron cytoplasmic protein 9.5, PGP 9.5, PGP9.5, Ubiquitin thioesterase L1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 11,400.00

CZK 11,400.00


Estimated to ship onMay 05, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1664

Request a Bulk Order

Select a Size

Change View
100 μL
CZK 11,400.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,400.00


Estimated to ship onMay 05, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1664

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MCA-BH7, monoclonal

species reactivity

human, bovine, rat

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)

General description

Ubiquitin carboxyl-terminal hydrolase isozyme L1 (UCH-L1), a thiol protease belonging to the peptidase C12 family, is involved in the processing of ubiquitin precursors and of ubiquitinated proteins. UCH-L1 recognizes and hydrolyzes peptide bonds at the C-termial glycine of ubiquitin. UCH-L1 is expressed in neurons and cells of the diffuse neuroendocrine system and their tumors. Defects in UCH-L1 are the cause of Parkinson disease type 5.

Immunogen

His-tagged recombinant protein corresponding to human UCHL1.

Application

Detect UCH-L1 using this mouse monoclonal antibody, Anti-UCHL1 Antibody, clone MCA-BH7 validated for use in western blotting & Immunofluorescence.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected UCHL1 in 10 µg of human brain tissue lysate.

Immunofluorescence Analysis: A 1:2,000 dilution from a representative lot detected UCHL1 in rat brain spinal cord tissue (Gerry Shaw. EnCor Biotechnology, Inc., Gainesville, Florida).

Quality

Evaluated by Western Blotting in rat brain spinal cord tissue lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected UCHL1 in 10 µg of rat brain tissue lysate.

Target description

~25 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Rat brain spinal cord tissue lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service