Skip to Content
Merck
All Photos(2)

Key Documents

MABN371

Sigma-Aldrich

Anti-KChIP2 potassium channel subunit Antibody, clone 60/73

clone 60/73, from mouse

Synonym(s):

Kv channel-interacting protein 2, KChIP2, A-type potassium channel modulatory protein 2, Potassium channel-interacting protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,500.00

CZK 10,500.00


Estimated to ship onMay 06, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,500.00


Estimated to ship onMay 06, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

60/73, monoclonal

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNIP2(30819)

General description

KCHIP2, also known as Kv channel-interacting protein 2 (KChIP2), or A-type potassium channel modulatory protein 2, or Potassium channel-interacting protein 2, and encoded by the gene Kcnip2/Kchip2, is the regulatory subunit of the Kv4/D Shal-type voltage gated inactivating potassium channel. KCHIP2 modulates the activities of Kv4.2 and Kv4.3 potassium channels. KCHIP2 is localized to the cell membrane but is a peripheral membrane protein. KCHIP2 is expressed in heart, brain and lung. In brain, KCHIP2 is abundantly expressed in striatum, hippocampus, and olfactory bulb, but moderate to low expression is seen in cerebral cortex thalamus and hypothalamus. KCHIP2 has multiple isoforms in the different tissues. Isoform 1 is predominant in cerebral cortex, striatum and hippocampus, and all three isoforms are equally expressed in olfactory bulb, but in heart only isoform 3 predominates.

Specificity

This antibody does not cross react with rat KChIP1 and KChIP3 (Prof. J. Trimmer, University of California, Davis).

Immunogen

GST-tagged recombinant protein corresponding to rat KChIP2 potassium channel subunit.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected KChIP2 potassium channel subunit in human cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-KChIP2 potassium channel subunit Antibody, clone 60/73 is validated for use in western blotting & IHC for the detection of KChIP2 potassium channel subunit.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected KChIP2 potassium channel subunit in 10 µg of mouse brain tissue lysate.

Target description

~ 35 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service