Skip to Content
Merck
All Photos(1)

Key Documents

MABN1792

Sigma-Aldrich

Anti-Sortilin Antibody, clone F11

clone F11, from mouse

Synonym(s):

Sortilin, 100 kDa NT receptor, Glycoprotein 95, Gp95, Neurotensin receptor 3, NT3, NTR3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 11,800.00

CZK 11,800.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 11,800.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,800.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F11, monoclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SORT1(6272)

General description

Sortilin (UniProt Q99523; also known as 100 kDa NT receptor, Glycoprotein 95, Gp95, Neurotensin receptor 3, NT3, NTR3) is encoded by the SORT1 (also known as LDLCQ6, NT3) gene (Gene ID 6272) in human. Sortlin belongs to the family of sorting receptors that are characterized by the Vps10p domain, which creates a tunnel cavity with its ten-bladed β-propeller for the binding of soluble ligands. These family members also contain a short cytoplasmic tail that harbors recognition motifs for subcellular sorting adaptor proteins. Sortilin is reported to bind PCSK9 in the trans-Golgi network with high-affinity in HepG2 cells. Sort1 knockout mice exhibit low levels of PCSK9 in plasma and its subcellular localization in hepatocytes is also altered. Sortilin is also found to co-localize with amyloid precursor protein (APP) in neurites and it facilitates APP processing toward production of sAPPα, possibly by optimizing APP as substrate for α-secretase cleavage. Sortilin is initially produced with a signal (a.a. 1-33) and a propeptide (a.a. 34-77) sequence, the removal of which yields the mature receptor (a.a. 78-831) with a large luminal/extracellular (a.a. 78-555) portion, a transmembrane region(a.a. 756-778), and a short cytoplasmic tail (a.a.779-831). Like the other family members (MPRs and SorLA), sortilin resides both on the membrane of intracellular vesicles and on the plasma membrane. Altered sortilin expressions due to SORT1 genetic variations affect serum levels of low density lipoprotein cholesterol (LDL-C) and contribute to the LDL-C level quantitative trait locus 6 (LDLCQ6) and susceptibility to myocardial infarction.

Immunogen

Epitope: Extracellular domain
Recombinant protein corresponding to the extracellular domain of human Sortilin.

Application

Anti-Sortilin Antibody, clone F11 is an antibody against Sortilin Antibody for use in Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blotting and ELISA.
Immunocytochemistry Analysis: A representative lot detected cellular localization of endogenous sortilin in isolated hippocampal neurons from new born (P0) wild-type, but not Sort1−/− C57BL/6J mice (Gustafsen, C., et al. (2013). J Neurosci. 33(1):64-71).
Immunocytochemistry Analysis: Representative lots detected cellular localization of exogenously expressed sortilin in transfected HEK293 cells by fluorescent immunocytochemistry (Larsen, J.V., et al. (2014). Biochem J. 457(2):277-288; Gustafsen, C., et al. (2013). J Neurosci. 33(1):64-71; Larsen, J.V., et al. (2010). Mol. Cell. Biol. 30(17):4175-4187; Nielsen, M.S., et al. (2007). Mol. Cell. Biol. 27(19):6842-6851).
ELISA Analysis: A representative lot was employed as the detection antibody in sandwich ELISA. A positive correlation was found between PCSK9 and soluble sortilin in human serum samples from healthy individuals (Gustafsen, C., et al. (2014). Cell Metab. 19(2):310-318).
Western Blotting Analysis: A representative lot detected different distribution of Sortilin than SorLA among subcellular fractions obtained by gradient centrifugation (Nielsen, M.S., et al. (2007). Mol. Cell. Biol. 27(19):6842-6851).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Sortilin in human cerebral cortex tissue.

Target description

~91 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Christophe Currie et al.
Cancers, 14(8) (2022-04-24)
Sortilin (SORT1) receptor-mediated endocytosis functions were exploited for this new approach for effective and safe treatments of gynecological cancers. Here, high expression of SORT1 was found in >75% of the clinically annotated ovarian and endometrial tumors analyzed by immunohistochemistry. Therefore
Cyndia Charfi et al.
Frontiers in oncology, 11, 760787-760787 (2021-11-10)
Vasculogenic mimicry (VM) is defined as the formation of microvascular channels by genetically deregulated cancer cells and is often associated with high tumor grade and cancer therapy resistance. This microcirculation system, independent of endothelial cells, provides oxygen and nutrients to
Peter Loof Møller et al.
Frontiers in cardiovascular medicine, 8, 652584-652584 (2021-05-04)
Genetic variants in the genomic region containing SORT1 (encoding the protein sortilin) are strongly associated with cholesterol levels and the risk of coronary artery disease (CAD). Circulating sortilin has therefore been proposed as a potential biomarker for cardiovascular disease. Multiple
Peder Madsen et al.
Scientific reports, 9(1), 611-611 (2019-01-27)
SorLA and Sortilin are multifunctional receptors involved in endocytosis and intracellular sorting of different and unrelated ligands. SorLA has recently attracted much attention as a novel strong risk gene for Alzheimer's disease, and much effort is currently being put into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service