Skip to Content
Merck
All Photos(3)

Key Documents

MABN1616

Sigma-Aldrich

Anti-GADD153/DDIT3/CHOP Antibody, clone 5E5.3

clone 5E5.3, from mouse

Synonym(s):

DNA damage-inducible transcript 3 protein, CCAAT/enhancer-binding protein homologous protein, C/EBP-homologous protein 10, C/EBP zeta, CHOP, CHOP-10, DDIT-3, Growth arrest and DNA damage-inducible protein GADD153

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,200.00

CZK 10,200.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,200.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,200.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E5.3, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CEBPZ(10153)

General description

DNA damage-inducible transcript 3 protein (UniProt P35638; also known as CCAAT/enhancer-binding protein homologous protein, C/EBP-homologous protein 10, C/EBP zeta, CHOP, CHOP-10, DDIT-3, Growth arrest and DNA damage-inducible protein GADD153) is encoded by the DDIT3 (Also known as CEBPZ, CHOP, CHOP10, GADD153) gene in human (Gene ID 1649). CHOP is a proapoptotic unfold protein response (UPR) gene and a well established marker for ER stress and UPR activation. The accumulation of unfolded or misfolded proteins in the ER induces the activation of three main UPR pathways, the inositol requiring enzyme-1 (IRE1) pathway, the activating transcription factor 6 (ATF6) pathway, and the protein kinase-like ER kinase (PERK) pathway. The PERK pathway involves PERK auto-phosphorylation/activation, resulting in PERK phosphorylation of eIF2α, which in turn leads to increased ATF4 transcription and ATF4-mediated CHOP upregulation.

Specificity

Clone 5E5.3 targeted region is present in both human GADD153/DDIT3/CHOP spliced isoforms reported by UniProt (P35638).

Immunogen

Epitope: Internal.
GST-tagged recombinant human GADD153/DDIT3/CHOP internal fragment.

Application

Immunocytochemistry Analysis: 5.0 µg/mL from a representative lot detected GADD153/DDIT3 in HeLa cells.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected GADD153/DDIT3 in human brain, kidney, and skeletal muscle tissue sections.
Research Category
Neuroscience
Research Sub Category
Apoptosis - Additional
This Anti-GADD153 Antibody, clone 5E5.3 is validated for use in Western Blotting, Immunocytochemistry, Immunohistochemistry (Paraffin) for the detection of CHOP.

Quality

Evaluated by Western Blotting Analysis in HUVEC lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected GADD153/DDIT3 in 10 µg of HUVEC lysate.

Target description

~20 kDa observed. 19.18 kDa (isoform 1) and 21.70 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service