Skip to Content
Merck
All Photos(1)

Key Documents

MABN139

Sigma-Aldrich

Anti-mGluR5 Antibody, clone 11D9.1

clone 11D9.1, from mouse

Synonym(s):

Metabotropic glutamate receptor 5, mGluR5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 11,100.00

CZK 11,100.00


Estimated to ship onApril 14, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1270

Request a Bulk Order

Select a Size

Change View
100 μG
CZK 11,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,100.00


Estimated to ship onApril 14, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1270

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11D9.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM5(2915)

General description

Metabotropic glutamate receptor 5 (mGluR5) is a member of the G-protein coupled receptor family and functions as a receptor for glutamate. mGluR5 activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current.

Immunogen

GST-tagged recombinant protein corresponding to human mGluR5.

Application

Detect mGluR5 using this mouse monoclonal antibody, Anti-mGluR5 Antibody, clone 11D9.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected mGluR5 in normal human brain tissue.

Quality

Evaluated by Western Blotting in rat brain micropreparation tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected mGluR5 in 10 µg of rat brain micropreparation tissue lysate. The lysate was not boiled during preperation for western blotting.

Target description

~150 kDa observed. The calculated molecular weight is 130 kDa, however mGluR5 has been shown as a ~150 kDa band in western blots (Romano, C., et al. (1996). J Biol Chem. 271(45):28612-28616).

Physical form

Format: Purified

Analysis Note

Control
Rat brain micropreparation tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly Smart et al.
Journal of psychiatry & neuroscience : JPN, 46(1), E1-E13 (2020-06-20)
Glutamate transmission is implicated in drug-induced behavioural sensitization and the associated long-lasting increases in mesolimbic output. Metabotropic glutamate type 5 (mGlu5) receptors might be particularly important, but most details are poorly understood. We first assessed in mice (n = 51

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service