Skip to Content
Merck

Skip To

MABF730

Anti-Foxp3 Antibody (mouse), APC, clone 3G3

clone 3G3, 0.2 mg/mL, from mouse

Synonym(s):

Forkhead box protein P3, Scurfin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg

Estimated to ship onJune 15, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 4,290.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
Allophycocyanin conjugate
Clone:
3G3, monoclonal
Application:
FACS
Citations:
-

CZK 4,290.00


Estimated to ship onJune 15, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

Allophycocyanin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

3G3, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Foxp3(20371)

General description

The 3G3 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is expressed at constitutively high levels in Treg cells, which are further identified as being CD4+ CD25+. In resting conventional T cells (CD4+ CD25-) Foxp3 expression is restricted, and upon TCR activation is expressed only transiently and in a small proportion of cells. However, the growth factor TGF-beta has been shown to induce expression of Foxp3 in naïve T cells, driving their development into Foxp3+ Tregs, which are called “induced” or “adaptive” Tregs. These cells are phenotypically similar to so-called “natural” Tregs (CD4+ CD25high Foxp3+) which originate in the thymus and comprise the majority of Treg cells. Tregs are critical for maintaining peripheral tolerance and are implicated in the development of autoimmunity.It is important to review the literature in choosing an antibody for the Foxp3 antigen in flow cytometry, as the potential for high background or non-specific staining may be observed. The 3G3 antibody may be used for intracellular detection of Foxp3 in cells from mouse and Rhesus macaque.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal antibody Anti-Foxp3 (mouse), APC & clone 3G3 validated for use in Flow Cytometry for the detection of Foxp3.

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Preparation Note

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Analysis Note

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg of this antibody detected Foxp3 in one million C57Bl/6 splenocytes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF462MABF732MABF464
conjugate

Allophycocyanin conjugate

conjugate

Allophycocyanin conjugate

conjugate

PE

conjugate

PE

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

Gene Information

mouse ... Foxp3(20371)

Gene Information

mouse ... Foxp3(20371)

Gene Information

mouse ... Foxp3(20371)

Gene Information

mouse ... Foxp3(20371)

clone

3G3, monoclonal

clone

3G3, monoclonal

clone

3G3, monoclonal

clone

3G3, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters