Skip to Content
Merck
All Photos(1)

Key Documents

MABF431

Sigma-Aldrich

Anti-CD45.1 Antibody (mouse), PerCP-Cy5.5, clone A20

clone A20, 0.2 mg/mL, from mouse

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,450.00

CZK 9,450.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 9,450.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

CZK 9,450.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PerCP-Cy5.5

antibody form

purified antibody

antibody product type

primary antibodies

clone

A20, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

Related Categories

General description

The A20 antibody reacts with mouse CD45.1, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal antibody Anti-CD45.1 (mouse), PerCP-Cy5.5, clone A20 validated for use in IP, IHC & Flow Cytometry for the detection of CD45.1.

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 µg of this antibody detected CD45.1 in one million C57Bl/6 or SJL splenocytes.

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service