Skip to Content
Merck

Skip To

MABF384

Anti-CD127 (IL-7Ra) Antibody (mouse), clone A7R34

clone A7R34, 0.5 mg/mL, from rat

Synonym(s):

Interleukin-17 receptor A, IL-17 receptor A, IL-17RA, CD217

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 7,820.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
A7R34, monoclonal
Application:
FACS, IF, IHC, IP
Citations:
1

CZK 7,820.00


Estimated to ship onJune 05, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

A7R34, monoclonal

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Il7R(16197)

General description

The A7R34 antibody is specific for mouse CD127, a 60-90 kDa cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cell development, allergy and autoimmunity.The A7R34 antibody may be used as a phenotypic marker for CD127 on immature B cells, on subsets of thymocytes which are double negative (CD4-CD8- ) or single positive (CD4+ or CD8+), and at low levels on mature, peripheral T cells. CD127 is a key marker, when used in combination with CD4 and CD25, to distinguish Treg and effector/memory Treg populations known as T(REM).

Application

This Rat Monoclonal Antibody Anti-CD127 (IL-7Ra) (mouse), clone A7R34, Cat. No. MABF384 is validated for use in IP, IHC, Immunofluorescence and Flow Cytometry for the detection of CD127 (IL-7Ra).

Physical form

Format: Purified

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF378MABF383MABF541
clone

A7R34, monoclonal

clone

A7R34, monoclonal

clone

A7R34, monoclonal

clone

A7R34, monoclonal

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

biological source

rat

biological source

rat

biological source

rat

biological source

rat

Gene Information

mouse ... Il7R(16197)

Gene Information

mouse ... Il7R(16197)

Gene Information

mouse ... Il7R(16197)

Gene Information

mouse ... Il7R(16197)

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

conjugate

unconjugated

conjugate

Allophycocyanin conjugate

conjugate

PerCP-Cy5.5

conjugate

PerCP-Cy5.5


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters