Skip to Content
Merck
All Photos(1)

Key Documents

MABF34

Sigma-Aldrich

Anti-Thy-1 Antibody, clone M5/49

clone M549.4.1, from rat

Synonym(s):

Thy-1 cell surface antigen, Thy-1 T-cell antigen, Thy-1 membrane glycoprotein, Thy-1 antigen, CD90 antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,740.00

CZK 9,740.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 9,740.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 9,740.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M549.4.1, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THY1(7070)

General description

Thy-1 membrane glycoprotein (Thy-1) is a glycosylphosphatidylinositol (GPI)-anchored surface protein belonging to the immunoglobulin superfamily and is a major cell surface protein found on both T–cells and in the brain. Thy-1 is thought to be involved in interactions taking place during synaptogenesis and other neurological events between cells or cells and their ligands. There are two known alleles for Thy-1; Thy1.1 and Thy1.2. Studies have shown N-glycosolation to take place at three locations on the protein.

Immunogen

B6 Anti-BNL4Φ lymphocytes

Application

Anti-Thy-1 Antibody, clone M5/49 detects level of Thy-1 & has been published & validated for use in WB.
Functional Assay: A previous lot of this antibody was used for the inhibition of B6 Anti-BNLΦ CTL mediated cell death. (Davignon, D., et al. (1981). PNAS USA. 78(7):4535-4539.)
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Neural Stem Cells

Quality

Evaluated by Western Blot in P388D1 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Thy-1 in 10 µg of P388D1 cell lysate.

Target description

~22 kDa observed.
Higher bands may be observed due to glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
P388D1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service