Skip to Content
Merck

Skip To

MABF3243

Anti-CD1d Antibody, clone CD1d42

Synonym(s):

Antigen-presenting glycoprotein CD1d, R3G1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 6,570.00
100 μg

Estimated to ship onJune 10, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 9,000.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
CD1d42, monoclonal
Application:
FACS, inhibition assay
Citations:
-

CZK 6,570.00


Estimated to ship onJune 08, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CD1d42, monoclonal

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable, inhibition assay: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD1D(912)

General description

Antigen-presenting glycoprotein CD1d (UniProt: P15813; also known as R3G1, CD1d) is encoded by the CD1D gene (Gene ID: 912) in human. Cd1d is a single-pass type 1 membrane glycoprotein, which is synthesized with a signal peptide (aa 1-19) that is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 20-301), a transmembrane domain (aa 302-322), and a short cytoplasmic domain (aa 323-335). It is expressed at high levels by human cortical thymocytes, and its expression is also observed on certain T cell leukemias. It serves as a key antigen-presenting molecule involved in the selection and activation of NK cells. It is a major-histocompatibility-complex (MHC) class-I-like molecule that presents self-lipid and non-self-lipid antigens to invariant natural killer T (iNKT) cells. Following engagement of the invariant T cell receptor (iTCR) by CD1d lipid complexes, iNKT cells proliferate, produce cytokines, and become cytotoxic, regulating innate and adaptive immune responses. (Ref.: Oleinika, K., et al. (2018). Nat. Commun. 9(1); Article 684; Spada, FM., et al. (2000). Eur. J. Immunol. 30; 3468-3477; Exley, M., et al. (2000). Immunology. 100(1): 37-47).

Immunogen

Recombinant fusion protein composed of 1, 2 and 3 domains of human CD1d fused to the Fc portion of murine IgG2a.

Application

Quality Control Testing

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD1d in one million Human peripheral blood mononuclear cells (PBMC).

Tested Applications

Inhibition Assay: A representative lot inhibited the responses of NK T cell clones to -GalCer in the presence of monocyte lineage APC. (Spada, F.M., et al. (2000). Eur J Immunol. 30(12):3468-77).

Flow Cytometry Analysis: A representative lot detected CD1d in Flow Cytometry (Spada, F.M., et al. (2000). Eur J Immunol. 30(12):3468-77).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone CD1d42 is a mouse monoclonal antibody that detects CD1d. It targets an epitope within the extracellular domain.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC983SAB1402712MABF165A
antibody form

purified antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

clone

CD1d42, monoclonal

clone

D5, monoclonal

clone

2E4, monoclonal

clone

SJ25C1, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

PE

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... CD1D(912)

Gene Information

human ... CD1D(912)

Gene Information

human ... CD1B(910)

Gene Information

human ... CD19(930)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters