Skip to Content
Merck
All Photos(1)

Documents

MABF2800

Sigma-Aldrich

Anti-EBNA1 Antibody, clone 1H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H4, monoclonal

mol wt

calculated mol wt 56.43 kDa
observed mol wt ~60 kDa

purified by

using protein G

species reactivity

Epstein-Barr virus

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

Epstein-Barr virus ... EBNA1(3783774)

Related Categories

General description

Epstein-Barr nuclear antigen 1 (UniProt: P03211; also known as EBNA-1, EBV nuclear antigen 1) is encoded by the EBNA1 (also known as BKRF1) gene (Gene ID: 3783709) in Epstein-Barr virus (EBV). EBV is a gamma-herpesvirus, a causative agent of infectious mononucleosis. Although the majority of virus infected individuals remain healthy, EBV has been implicated in the pathogenesis of African Burkitt s lymphoma and nasopharyngeal carcinoma. EBNA1, a multi-functional protein, plays an important role in maintaining EBV in the host cell and has been implicated in host cell immortalization. It plays an essential role in replication and partitioning of viral genomic DNA during latent viral infection. During this phase, the circular double-stranded viral DNA undergoes replication once per cell cycle and is efficiently partitioned to the daughter cells. EBNA1 activates the initiation of viral DNA replication through binding to specific sites in the viral latent origin of replication, oriP. It is also reported to direct the segregation of viral episomes by mediating their attachment to host cell metaphase chromosomes and is known to activate the transcription of several viral latency genes. By counteracting the stabilization of host p53, it also reduces apoptosis and increases host cell survival. EBNA1 is also a risk factor in a variety of malignancies and is the only viral protein found in all EBV-related malignancies. Its C-terminal region contains a dimerization domain and a DNA binding domain (aa 459-607). Although this domain is essential for its replication function of EBNA1, it is not sufficient for this activity and the N-terminal half of EBNA1 is also required.

Specificity

Clone 1H4 is a rat monoclonal antibody that detects Epstein-Barr nuclear antigen 1.

Immunogen

Lambda-cro/lacZ-EBNAl-fusion protein s emulsified with complete Freund s adjuvant.

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected EBNA-1 in Raji cell lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected EBNA1 in Western Blotting applications (Grasser, F.A., et al. (1994). Blood. 84(11):3792-8; Humme, S., et al. (2003). Proc Natl Acad Sci USA. 100(19):10989-94; AlQarni, S., et al. (2018). Oncogene. 37(29):3998-4012).

Immunohistochemistry Analysis: A representative lot detected EBNA1 in Immunohistochemistry applications (Grasser, F.A., et al. (1994). Blood. 84(11):3792-8; Herrmann, K., et al. (2003). Breast Cancer Res. 5(1):R13-7; Chatterjee, B., et al. (2019). PLoS Pathog. 15(5):e1007748).

Immunoprecipitation Analysis: A representative lot detected EBNA1 in Immunoprecipitation applications (Schepers, A., et al. (2001). EMBO J. 20(16):4588-602).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-EBNA1, clone 1H4, Cat. No. MABF2800, is a rat monoclonal antibody that detects EBNA-1 and is tested for use in Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service