Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 11,100.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
AB34_41.4H12.4A7, monoclonal
species reactivity
human
technique(s)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... IL1B(3553)
General description
Immunogen
Application
Inflammation & Immunology
Cytokines & Cytokine Receptors
Physical form
Preparation Note
Analysis Note
Human Interleukin-1 beta recombinant protein
Western Blot Analysis: 0.5 µg/mL of this antibody detected 10 µg of human Interleukin-1 beta recombinant protein.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information human ... IL1B(3553) | Gene Information human ... IL1B(3553) | Gene Information human ... IL1B(3553) | Gene Information human ... IL1B(3553) |
| clone AB34_41.4H12.4A7, monoclonal | clone polyclonal | clone polyclonal | clone 2A8, monoclonal |
| species reactivity human | species reactivity human | species reactivity human, rat, mouse | species reactivity human |
| biological source mouse | biological source rabbit | biological source rabbit | biological source mouse |
| antibody form purified antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



