Skip to Content
Merck
All Photos(1)

Key Documents

MABF1773

Sigma-Aldrich

Rat IgG1 Negative Control, clone HRPN, APC conjugate Antibody

clone HRPN, from rat, purified by affinity chromatography

Synonym(s):

Immunoglobulin 1, IgG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rat

Quality Level

antibody form

affinity isolated antibody

clone

HRPN, monoclonal

purified by

affinity chromatography

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Application

This rat monoclonal Rat IgG1 Negative Control, clone HRPN, APC conjugate, Cat. No. MABF1773 is validated for use in Flow Cytometry.

Other Notes

Concentration: Please refer to lot specific datasheet.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service