Skip to Content
Merck
All Photos(1)

Key Documents

MABF145A

Sigma-Aldrich

Anti-CD90.2 (mouse), violetFluor® 450, clone 30-H12 Antibody

clone 30-H12, from rat, violetFluor® 450

Synonym(s):

Thy-1 membrane glycoprotein, Thy-1 antigen, CD90, CD90.2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CZK 3,890.00
100 μG
CZK 7,690.00

CZK 3,890.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
CZK 3,890.00
100 μG
CZK 7,690.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

CZK 3,890.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

biological source

rat

Quality Level

conjugate

violetFluor® 450

antibody form

purified antibody

antibody product type

primary antibodies

clone

30-H12, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2b

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Thy1(21838)

General description

The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-CD90.2 (mouse), violetFluor® 450, clone 30-H12 Antibody is an antibody against CD90.2 for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.25 μg from a representative lot detected CD90.2 in one million C57Bl/6 splenocytes.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

violetFluor is a registered trademark of Tonbo Biotechnologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service