Skip to Content
Merck

Skip To

MABE988

Anti-Homeobox protein CDX-2 Antibody, clone 7D9.1

clone 7D9.1, from mouse

Synonym(s):

Homeobox protein CDX-2, Homeobox protein CDX-2, CDX-3, Caudal-type homeobox protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 19, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7D9.1, monoclonal
Application:
IHC, WB
Citations:
-

CZK 10,500.00


Estimated to ship onJune 19, 2026Details

A recombinant, preservative-free antibody is available for your target. Try ZRB2817

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDX2(1045)

General description

Homeobox protein CDX-2 (UniProt Q99626; also known as Caudal-type homeobox protein 2, Caudal type homeobox transcription factor 2, CDX-3) is encoded by the CDX2 (also known as CDX3, CDX2/AS) gene (Gene ID 1045) in human. CDX-2 is a transcription factor that plays an important role during embryonic development, where it determines intestinal identity in the presumptive gut endoderm. CDX-2 is also required for maintenance of intestinal identity in the intestinal epithelium throughout adulthood. Mitogen-activated protein kinases (MAPK) are reported to modulate CDX2 transcriptional activity and half-life via phosphorylation. Reduction of CDX2 expression facilitates tumor progression in murine models of colorectal cancer (CRC) and decreased CDX2 expression level in CRC correlates with poor prognosis in human.
~35/39 kDa observed. Target band often appears as a doublet due to posttranslational modifications (phosphorylations & polyubiquitinations).

Immunogen

Epitope: Internal
GST-tagged recombinant protein corresponding to human Homeobox protein CDX-2.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Homeobox protein CDX-2 in human colon and human colorectal cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Homeobox protein CDX-2 Antibody, clone 7D9.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Homeobox protein CDX-2.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human colorectal adenocarcinoma DLD-1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Homeobox protein CDX-2 in 10 µg of human colorectal adenocarcinoma DLD-1 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV31476HPA049580SAB4301787
biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... CDX2(1045)

Gene Information

human ... CDX2(1045)

Gene Information

human ... CDX2(1045)

Gene Information

human ... CDX2(1045)

clone

7D9.1, monoclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human

species reactivity

human, bovine, rabbit, mouse, rat, guinea pig, dog

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters