Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 15,200.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
12C2.1, monoclonal
species reactivity
human, mouse
technique(s)
immunohistochemistry: suitable (paraffin), western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... PHF1(5252)
General description
Immunogen
Application
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PHF1 in 10 µg of HeLa cell lysate.
Epigenetics & Nuclear Function
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PHF1 in 10 µg of mouse heart tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Gene Information human ... PHF1(5252) | Gene Information human ... PHF1(5252) | Gene Information human ... PHF12(57649) | Gene Information human ... PHF2(5253) |
| biological source mouse | biological source rabbit | biological source rabbit | biological source mouse |
| clone 12C2.1, monoclonal | clone polyclonal | clone polyclonal | clone 3E7, monoclonal |
| antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form ascites fluid |
| species reactivity human, mouse | species reactivity bovine, human, dog, horse | species reactivity human | species reactivity human |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



