Skip to Content
Merck

MABE628

Anti-CHUK, clone 3G12H9 Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

100 μL

CZK 11,200.00

CZK 11,200.00


Estimated to ship onMarch 20, 2026Details


Request a Bulk Order

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3G12H9, monoclonal
Application:
ICC, WB
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

unconjugated

Quality Level

antibody form

ascites fluid

clone

3G12H9, monoclonal

purified by

(unpurified)

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

target post-translational modification

unmodified

General description

Purified recombinant fragment of human CHUK expressed in E. Coli.
~75 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Application

Immunocytochemistry Analysis (ICC): A 1:200-1,000 dilution from a representative lot detected CHUK in NIH/3T3 cells.

Physical form

Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in Raji, Jurkat, THP-1 and K562 cell lysates.

Western Blotting Analysis (WB): A 1:500-2,000 dilution of this antibody detected CHUK in Raji, Jurkat, THP-1 and K562 cell lysates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
MABE62804055977171723

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service