Skip to Content
Merck
All Photos(6)

Key Documents

MABE461

Sigma-Aldrich

Anti-CRTC1 Antibody/TORC-1, clone 5F3.1

clone 5F3.1, from mouse

Synonym(s):

CREB-regulated transcription coactivator 1, Mucoepidermoid carcinoma translocated protein 1, Transducer of regulated cAMP response element-binding protein 1, TORC-1, Transducer of CREB protein 1, CRTC1/TORC-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,500.00

CZK 10,500.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,500.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5F3.1, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CRTC1(23373)

General description

CREB-regulated transcription coactivator 1 (UniProt Q6UUV9; also known as Mucoepidermoid carcinoma translocated protein 1, TORC-1, Transducer of CREB protein 1, Transducer of regulated cAMP response element-binding protein 1) is encoded by the CRTC1 (also known as KIAA0616, MECT1, TORC1, WAMTP1) gene (Gene ID 23373) in human. CRTC1 belongs to a family of glycosylated Ser/Thr-rich phosphoproteins that bind the basic leucine zipper (bZIP) region of Ca2+/cAMP-responsive element-binding protein (CREB) and function as CREB coactivators. CRTCs participate in CREB-mediated transcriptions of both canonical cellular cAMP response elements (CREs) and the non-canonical viral CREs. CRTC1, specifically, is involved in CREB-mediated transcription of various neuronal immediate early genes (IEGs) as well as CREB-dependent hepatitis B virus (HBV) replication.

Specificity

Expected to react with all three human spliced isoforms

Immunogen

Epitope: Internal (N-terminal half)
GST-tagged recombinant protein corresponding to human CRTC1/TORC-1 sequence within the N-terminal half..

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-CRTC1 Antibody/TORC-1, clone 5F3.1 is validated for use in Western Blotting, Immunohistochemistry, Immunoprecipitation for the detection of CRTC1.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected CRTC1/TORC-1 in 10 µg of mouse hypothalamus tissue lysate.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected CRTC1/TORC-1 in human cerebral cortex, rat cerebral cortex, human cerebellum, and rat cerebellum tissue.
Immunoprecipitation Analysis: A representative lot immunoprecipitated endogenous CRTC1/TORC-1 from mouse hippocampus tissue lysates, as well as exogenously expressed murine CRTC1/TORC-1, but not CRTC2/TORC-2, from lysates of transfected HEK293 cells (Courtesy of Dr. Carlos A. Saura , Universitat Autònoma de Barcelona, Spain).
Western Blotting Analysis: A representative lot detected endogenous CRTC1/TORC-1 in human, mouse, and rat hippocampus tissue lysates, as well as exogenously expressed murine CRTC1/TORC-1 in lysates from transfected HEK293 cells (Courtesy of Dr. Carlos A. Saura , Universitat Autònoma de Barcelona, Spain).

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CRTC1/TORC-1 in 10 µg of mouse cerebellum tissue lysate.

Target description

~68 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service