Skip to Content
Merck

Skip To

MABE325

Anti-p21WAF1 Antibody, clone EA10

clone EA10, from mouse

Synonym(s):

Cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, Melanoma differentiation-associated protein 6, MDA-6, p21

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
EA10, monoclonal
Application:
FACS, WB
Citations:
21

CZK 11,800.00


Estimated to ship onJune 03, 2026Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1141

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EA10, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDKN1A(1026)

General description

The tumor suppressor p53 transcriptionally activates a number of genes including the WAF1/CIP1 gene in response to DNA damage. The ~21 kDa product of the WAF1 gene is found in a complex involving cyclins, CDKs, and PCNA in normal cells but not transformed cells and appears to be a universal inhibitor of CDK activity. One consequence of p21WAF1 binding to and inhibiting CDKs is the prevention of CDK-dependent phosphorylation and subsequent inactivation of the Rb protein which is essential for cell cycle progression. p21WAF1 is, therefore, a potent and reversible inhibitor of cell cycle progression at both the G1 and G2 checkpoints, presumably to allow sufficient time for DNA repair to be completed. Irreversible G1 or G2 arrest leads to apoptosis. While the role of p21WAF1 in apoptosis is less clear, it is known that p53-mediated apoptosis leads to increased WAF1 expression. Induction of p21WAF1 can occur by both p53-dependent and p53-independent mechanisms, in response to certain observed conditions. p21WAF1 has also been identified as a gene involved in cellular senescence, termed sdi1. Its overexpression was observed to inhibit cellular growth.
~21 kDa observed

Immunogen

Recombinant protein corresponding to human p21WAF1.

Application

Anti-p21WAF1 Antibody, clone EA10 is a Mouse Monoclonal Antibody for detection of p21WAF1 & has been validated in WB, FC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot Analysis: A representative lot from an independent laboratory detected p21WAF1 in non-irradiated and gamma irradiated CLL cells (Carter, A., et al. (2004). Br J Haematol. 127(4):425-428).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected p21WAF1 in normal human lower back tissue (el-Deiry, W. S., et al. (1995). Cancer Res. 55(13):2910-2919.).

Flow Cytometry Analysis: A representative lot from an independent laboratory detected p21WAF1 in non-irradiated and gamma irradiated CLL cells (Carter, A., et al. (2004). Br J Haematol. 127(4):425-428).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HT-29 cell lysate
Evaluated by Western Blot in HT-29 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected p21WAF1 in 10 µg of HT-29 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1484SAB5700189OP64
antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

Gene Information

human ... CDKN1A(1026)

Gene Information

human ... CDKN1A(1026)
mouse ... Cdkn1a(12575)

Gene Information

human ... CDKN1A(1026)

Gene Information

human ... CDKN1A(1026)

clone

EA10, monoclonal

clone

CP74, monoclonal

clone

polyclonal

clone

EA10, monoclonal

species reactivity

human

species reactivity

mouse, human

species reactivity

rat, mouse, human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters