Skip to Content
Merck

Skip To

MABE1529

Anti-BRD7 Antibody, clone BRM 2D3

clone BRM203, from rat

Synonym(s):

BRD7, Bromodomain-containing protein 7, 75 kDa bromodomain protein, Protein CELTIX-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on16 November 2026fromAreál Kühne+Nagel spol. s r.o.

11 200,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
BRM203, monoclonal
Application:
IP, WB
Citations:
-

11 200,00 CZK


Estimated to ship on16 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BRM203, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... BRD7(29117)

General description

Bromodomain-containing protein 7 (UniProt Q9NPI1; also known as 75 kDa bromodomain protein, Protein CELTIX-1) is encoded by the BRD7 (also known as BP75, CELTIX1) gene (Gene ID 29117) in human. BRD7 is a ubiquitously expressed single bromodomain-/BRD-containing member of the family of BRD-containing proteins. BRD7 functions as a subunit of the SWI/SNF chromatin-remodeling complex involved in transcription regulation. It also interacts with the well known tumor suppressor p53 to trans-activate genes involved in cell cycle arrest. BRD7 is downregulated in colorectal carcinoma and nasopharyngeal carcinoma, and BRD7 overexpression is reported to inhibit nasopharyngeal carcinoma cell proliferation via cell cycle arrest by transcriptionally regulating ras/MEK/ERK, Rb/E2F, b-catenin and ERK pathways. BRD7 mRNA has been identified as a silencing target of miR-300, overexpression of miR-300 promotes the proliferation, invasion and EMT of osteosarcoma cells through targeting BRD7 mRNA.
~75 kDa observed. 74.14 kDa (isoform 1), 74.27 kDa (isoform 2) calculated. Added Uncharacterized band(s) may appear in some lysates.

Immunogen

Recombinant human BRD7.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-BRD7 Antibody, clone BRM 2D3 is validated for use in Western Blotting, Immunoprecipitation for the detection of BRD7.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected BRD7 in 10 µg of HEK293 and HeLa cell lysates.
Immunoprecipitation Analysis: A representative lot immunoprecipitated BRD7 from H1299 cell lysate (Courtesy of Thomas Dobner, Institut fur Medizinische Mikrobiologie und Hygiene, Universitat Regensburg, D-93053 Regensburg, Germany).

Biochem/physiol Actions

Clone BRM 2D3 detects human 75 kDa bromodomain protein, also known as protein CELTIX-1.

Physical form

Format: Purified
Protein G purified.
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected BRD7 in 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: MABE66

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200047MABE918ABE1391
description

clone BRM203, from rat

description

clone BRM 2D3, purified from hybridoma cell culture

description

clone 1F11, from mouse

description

serum, from rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

rat, mouse, monkey, human, bovine, canine

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

purified immunoglobulin

antibody form

serum

Gene Information

human ... BRD7(29117)

Gene Information

human ... BRD7(29117)

Gene Information

human ... BRD4(23476)

Gene Information

mouse ... Brd4(57261)

clone

BRM203, monoclonal

clone

BRM 2D3, monoclonal

clone

1F11, monoclonal

clone

polyclonal

biological source

rat

biological source

rat

biological source

mouse

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters