Skip to Content
Merck

Skip To

MABE148

Anti-HMGB1 Antibody, clone HMG1-5H6

clone HMG1-5H6, from hamster

Synonym(s):

High mobility group protein 1, Sulfoglucuronyl carbohydrate binding protein, high mobility group box 1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
13 100,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
HMG1-5H6, monoclonal
Application:
FACS, IF, WB
Citations:
1

13 100,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

hamster

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

HMG1-5H6, monoclonal

species reactivity

rat, mouse, human

technique(s)

flow cytometry: suitable, immunofluorescence: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)

General description

HMG-1 (High-Mobility-Group protein, amphoterin) is a highly conserved protein with more than 95% amino acid identity between rodent and human HMG-1. It is a prevalent non-histone chromatin component and a non-sequence specific DNA binding protein. HMG-1 consists of two homologous HMG boxes rich in basic amino acids and an acidic tail at the carboxy-terminus. HMG-1 is involved in the regulation of chromatin structure as well as being involved, either as positive or negative factors with various aspects of DNA replication, transcription, repair, and ligation. HMG-1, identified as a membrane associated protein termed “amphoterin,” mediates neurite outgrowth, tumor outgrowth, and metastasis. It participates in plasminogen activation and is recognized as a late mediator of endotoxin lethality in mice.
~ 30 kDa observed

Immunogen

Recombinant protein corresponding to mouse HMGB1.

Application

Immunofluorescence Analysis: 10 µg/mL from a representative lot detected HMGB1 in NIH/3T3 cells. (Image courtesy of Dr. Hideo Yagita, Juntendo University School of Medicine.)

Flow Cytometry Analysis: A representative lot detected HMGB1 in RAW264.7 cells treated with Cytofix/Cytoperm. (Image courtesy of Dr. Hideo Yagita, Juntendo University School of Medicine.)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Cell Cycle, DNA Replication & Repair
Use Anti-HMGB1 Antibody, clone HMG1-5H6 (Hamster Monoclonal Antibody) validated in WB, IF, FC to detect HMGB1 also known as High-mobility group box 1.

Physical form

Format: Purified
Protein G
Purified hamster monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate
Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 1:500 dilution of this antibody detected HMGB1 on 10 µg of NIH/3T3 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0003146M8H9539H9664
description

clone HMG1-5H6, from hamster

description

clone 2F6, purified immunoglobulin, buffered aqueous solution

description

affinity isolated antibody, buffered aqueous solution

description

affinity isolated antibody, buffered aqueous solution

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human, rat, mouse

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

HMG1-5H6, monoclonal

clone

2F6, monoclonal

clone

polyclonal

clone

polyclonal

biological source

hamster

biological source

mouse

biological source

rabbit

biological source

rabbit

Gene Information

human ... HMGB1(3146)

Gene Information

human ... HMGB1(3146)

Gene Information

human ... HMGB1(3146)
mouse ... Hmgb1(15289)
rat ... Hmgb1(25459)

Gene Information

human ... HMGB1(3146)
mouse ... Hmgb1(15289)
rat ... Hmgb1(25459)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters