Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 10,500.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4B1-F6, monoclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... AGO3(192669)
General description
Ago3 is indicated to play a role in the RNA interference (RNAi) pathway, which is important for heterochromatin formation and accurate chromosome segregation. Unlike Ago1 and Ago2, Ago3 and AG04 do not possess the ability to dissociate miRNA duplexes in a minimal in vitro system. Re-introduction of any Ago variant into embyronic (ES) cells deficient for expression of all four Ago variants will rescue miRNA silencing defects and reduce apoptosis.
Immunogen
Application
Physical form
Analysis Note
Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Ago3 in HeLa cells.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified immunoglobulin | antibody form purified from hybridoma cell culture | antibody form purified immunoglobulin | antibody form purified antibody |
| biological source mouse | biological source mouse | biological source mouse | biological source rat |
| Gene Information human ... AGO3(192669) | Gene Information human ... AGO2(27161) | Gene Information human ... AGO1(26523) | Gene Information human ... AGO2(27161) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| clone 4B1-F6, monoclonal | clone AGO2-10, monoclonal | clone 2A8, monoclonal | clone 11A9, monoclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



