Skip to Content
Merck

Skip To

MABE1121

Anti-Ago1 Antibody, clone 4B8

clone 4B8, from rat

Synonym(s):

Protein argonaute-1, Argonaute1, hAgo1, Argonaute RISC catalytic component 1, Eukaryotic translation initiation factor 2C 1, eIF-2C 1, eIF2C 1, Putative RNA-binding protein Q99, Ago1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4B8, monoclonal
Application:
WB
Citations:
3

CZK 10,500.00


Available to ship TODAYDetails

A recombinant, preservative-free antibody is available for your target. Try ZRB2132

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B8, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AGO1(26523)

General description

Protein argonaute-1 (UniProt Q9UL18; also known as Argonaute1, Argonaute RISC catalytic component 1, eIF-2C 1, eIF2C 1, Eukaryotic translation initiation factor 2C 1, hAgo1, Putative RNA-binding protein Q99) is encoded by the AGO1 (also known as EIF2C, EIF2C1) gene (Gene ID 26523) in human. Ago1 belongs to the family of argonaute proteins involved in gene expression by regulating gene transcription and RNA splicing in the nucleus, as well as by regulating posttranscriptional gene silencing mediated by siRNAs and microRNAs in the cytosol. Ago1 is shown to interact with chromatin modifiers, splicing factors, and RNA polymerase II. Ago1 and the related Ago2 are also shown to target Hepatitis C Virus (HCV) RNÁ 5′-untranslated region (5′-UTR) in an miR-122-dependent manner, resulting in viral RNA stability and translation stimulation.
~97 kDa observed

Immunogen

GST-tagged recombinant human Ago1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Ago1 in 10 µg of K562 cell lysate.
Western Blotting Analysis: A representative lot detected Ago1-containing messenager ribonucleoprotein (mRNP) complexes primarily in the low density fractions of sucrose gradient-fractionated HEK293 lysate (Höck, J., et al. (2007). EMBO Rep. 8(11):1052-1060).
Immunoprecipitation Analysis: Representative lots immunoprecipitated FLAG-tagged Ago1, but not FLAG-tagged Ago2, Ago3, or Ago4 (Ender, C., et al. (2008). Mol. Cell. 32(4):519-528; Beitzinger, M., et al. (2007). RNA Biol. 4(2):76-84).
RNA Binding Protein Immunoprecipitation (RIP): Representative lots co-immunoprecipitated Ago1-associated RNAs, including mRNAs, miRNAs, and snoRNAs from human cell lysates (Li, Z., et al. (2009). J. Virol. 83(24):12751-12758; Ender, C., et al. (2008). Mol. Cell. 32(4):519-528; Beitzinger, M., et al. (2007). RNA Biol. 4(2):76-84).

Biochem/physiol Actions

Clone 4B8 sepcifically immunoprecipitated FLAG-tagged Ago1,but not FLAG-tagged Ago2, Ago3, or Ago4 (Ender, C., et al. (2008). Mol. Cell. 32(4):519-528).

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Ago1 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE143SAB4200084SAB1303264
clone

4B8, monoclonal

clone

4G7-E12, monoclonal

clone

4B8, monoclonal

clone

polyclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

IgG fraction of antiserum

species reactivity

human

species reactivity

human

species reactivity

human, canine, mouse, monkey, bovine

species reactivity

human

biological source

rat

biological source

mouse

biological source

rat

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... AGO1(26523)

Gene Information

human ... AGO1(26523)

Gene Information

human ... EIF2C1(26523)
mouse ... Eif2c1(236511)

Gene Information

human ... AGO2(27161)
mouse ... Ago2(239528)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters