Skip to Content
Merck
All Photos(2)

Documents

MABD50

Sigma-Aldrich

Anti-MyT1L Antibody, clone 20B4.1

clone 20B4.1, from mouse

Synonym(s):

Myelin transcription factor 1-like protein, MyT1-L, MyT1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20B4.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYT1L(23040)

General description

Myelin transcription factor 1-like (Myt1L) is a zinc finger protein belonging to the myelin transcription factor 1 family. It influences the expression of genes involved in the proliferation and differentiation of oligodentrocytes in the developing central nervous system. Myt1L may contribute to the silencing of its target genes by recruiting Sin3B that binds to and enhances the activity of HDAC. Several studies have implicated Myt1L in the pathogenesis of complex psychiatric diseases such as schizophrenia, major depressive disorder, and bipolar disorder.

Immunogen

GST-tagged recombinant protein corresponding to human MyT1L.

Application

Detect the MyT1L protein using this Anti-MyT1L, clone 20B4.1 validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:10-1:50 dilution from a representative lot detected MyT1L in human pons, mouse frontal cortex, and mouse hippocampus tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse E17 spinal cord tissue lysate.

Western Blot Analysis: A 1:1.000 dilution of this antibody detected MyT1L on 10 µg of mouse E17 spinal cord tissue lysate.

Target description

~170 kDa observed. Uniprot describes three isoforms at 21, 126, and 133 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse E17 spinal cord tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service