Skip to Content
Merck
All Photos(1)

Key Documents

MABC959

Sigma-Aldrich

Anti-CD1d Antibody, clone WTH-1

clone WTH-1, from mouse

Synonym(s):

Antigen-presenting glycoprotein CD1d, CD1d

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,900.00

CZK 12,900.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 12,900.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 12,900.00


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

WTH-1, monoclonal

species reactivity

mouse, rat

technique(s)

activity assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Antigen-presenting glycoprotein CD1d (UniProt Q63493; also known as CD1D antigen, CD1d1 antigen, T-cell surface glycoprotein CD1d) is encoded by the Cd1d (also known as Cd1d1) gene (Gene ID 25109) in rat species. CD1d is a nonpolymorphic MHC class I-like molecule which presents nonpeptide ligands (e.g. glycolipids) to natrual killer T (NKT) cells. NKT cells are known to have multiple effects on innate and adaptive immune responses and on the development of pathological conditions. In additon to hematopoietic cells, CD1d protein is also reported to be detected in non-lymphatic tissues in rat, such as the exocrine pancreas and Paneth cells. CD1d is initially produced with a signal peptide sequence (a.a. 1-17), the removal of which yields the mature CD1d with a large extracellular region (a.a.18-304), a transmembrane domain (a.a. 305-325), and a short cytoplasmic tail (a.a. 326-336).

Application

This Anti-CD1d Antibody, clone WTH-1 is validated for use in Flow Cytometry, Western Blotting, Immunoprecipitation, Immunohistochemistry and Activity Assay for the detection of CD1d.
Western Blotting Analysis: A representative lot detected endogenous CD1d in rat spleen, thymus, and pancreas tissue homogenates, as well as exogenously expressed murine and rat CD1d in lysates of transfected Raji cells (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Flow Cytometry Analysis: A representative lot detected the surface expression of CD1d on isolated thymocytes from rats and mice, as well as on Raji transfectants expressing murine or rat CD1d (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Immunoprecipitation Analysis: A representative lot immunoprecipitated CD1d from isolated mouse and rat thymocytes, as well as exogenously expressed murine and rat CD1d from transfected Raji cells (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Immunohistochemistry Analysis: A representative lot detected CD1d immunoreactivity using frozen tissue sections from rats and mice (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Activity Assay: A representative lot inhibited CD1d antigen presentation cells from activating type I NK cells using total splenocytes from rats and mice. Clone WTH-1 also blocked mCD1d1-transduced LBB cells from activating co-cultured VIII24.1 murine type II NKT cells, while WTH-1 enhanced XV19.2 murine type II NKT cell activation by mCD1d1-transduced Raji cells (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Application Note: Clone WTH-1 (Cat. No. MABC959) and WTH-2 (Cat. No. MABC960) gave inferior IHC staining in mouse than in rat organs, especially in non-lymphatic tissues. In addition, clone WTH-2 produced a much lower signal in mice than mAb WTH-1. To detect low-grade expression, mouse non-lymphatic organs should be stained using a highly sensitive tyramide amplification procedure with inclusion of C57BL/6 CD1d2/2 mouse organs as negative controls (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).

Quality

Evaluated by Flow Cytometry in rat thymocytes and splenocytes.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD1d in rat thymocytes and splenocytes.

Target description

~38 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service