Skip to Content
Merck
All Photos(1)

Key Documents

MABC664

Sigma-Aldrich

Anti-CLEC2A Antibody, clone 8D2.1

clone 8D2.1, from mouse

Synonym(s):

C-type lectin domain family 2 member A, Keratinocyte-associated C-type lectin, KACL, Proliferation-induced lymphocyte-associated receptor, PILAR, CLEC2A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 11,100.00

CZK 11,100.00


Estimated to ship onMarch 26, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 11,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,100.00


Estimated to ship onMarch 26, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8D2.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLEC2A(387836)

General description

C-type lectin domain family 2 member A (CLEC2A) is also called proliferation-induced lymphocyte-associated receptor (PILAR) and Keratinocyte-associated C-type lectin (KACL). CLEC2A is involved in modulating T-cell expansion. It is a 32 kDa single-pass type II membrane protein that belongs to the CTL/CTLD superfamily. CLEC2A is mainly expressed in skin and highly expressed in CD8(+), B lymphocytes and naive CD4(+) T cells. Modulation of CLEC2A signaling may be important for treatment of autoimmune diseases and cancer. CLEC2A is associated with expansion of TCR-stimulated T-cells, modulates the capacity of T-cells to home to lymph nodes through SELL, and facilitates immune recognition of keratinocytes by stimulating natural killer cell mediated cytotoxicity.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human CLEC2A.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-CLEC2A Antibody, clone 8D2.1 is validated for use in Western Blotting for the detection of CLEC2A.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CLEC2A in 200 µg of Raji cell lysate.

Target description

~18 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service