Skip to Content
Merck

Skip To

MABC598

Anti-PACS2 Antibody, clone 9F12.2

clone 9F12.2, from mouse

Synonym(s):

Phosphofurin acidic cluster sorting protein 2, PACS1-like protein, PACS2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

11 100,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9F12.2, monoclonal
Application:
WB
Citations:
-

11 100,00 CZK


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F12.2, monoclonal

species reactivity

mouse, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pacs2(217893)

General description

Phosphofurin acidic cluster sorting protein 2 (PACS2), also known as PACS-2, PACS1-like protein, and encoded by the gene name PACS2/ KIAA0602/ PACS1L, is a amember of the PACS family. PACS2 is a multifunctional sorting protein that controls the endoplasmic reticulum (ER)-mitochondria communication, including the apposition of mitochondria with the ER and ER homeostasis. Additionally, PACS2 translocates BIB to mitochondria in response to apoptic inducers, which initiates a sequence of events including the formation of mitochondrial truncated BID, the release of cytochrome C, and the activation of caspase-3 thereby causing cell death. Recent studies have indicated that PACS2 may also be involved in ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments. PACS2 interacts with BID, PKD2, and HIV-1 Nef.
~100 kDa observed

Immunogen

GST-tagged recombinant protein corresponding to Human PACS2.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-PACS2 antibody is validated for use in Western Blotting for the detection of PACS2.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected PACS2 in 10 µg of mouse hypothalamus tissue.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat hippocampus tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PACS2 in 10 µg of rat hippocampus tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2105784HPA001423HPA038914
description

clone 9F12.2, from mouse

description

affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Gene Information

mouse ... Pacs2(217893)

Gene Information

human ... PACS2(23241)

Gene Information

human ... PACS2(23241)

Gene Information

human ... PACS1(55690)

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

9F12.2, monoclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse, rat

species reactivity

guinea pig, horse, bovine, human, rat, dog, mouse

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters