Skip to Content
Merck

MABC565

Anti-SPOP Antibody, clone 9B7.1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9B7.1, monoclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

antibody form

purified antibody

clone

9B7.1, monoclonal

purified by

using protein G

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

General description

GST-tagged recombinant protein corresponding to human SPOP.
~42 kDa observed. Uncharacterized band(s) may appear in some lysates.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected SPOP in human liver and human kidney tissue.

Physical form

Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SPOP in 10 µg of HeLa cell lysate.

Other Notes

Human. Predicted to react with Rat, Mouse based on 100% sequence homology. Other homologies: 100% homologus to mouse and rat..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library