Skip to Content
Merck
All Photos(2)

Key Documents

MABC543

Sigma-Aldrich

Anti-FBPase 1, clone 13A8.1 Antibody

clone 13A8.1, from mouse

Synonym(s):

Fructose-1,6-bisphosphatase 1, FBPase 1, D-fructose-1,6-bisphosphate 1-phosphohydrolase 1, Liver FBPase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 11,100.00

CZK 11,100.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
CZK 11,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,100.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13A8.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FBP1(2203)

General description

Fructose-1, 6-bisphosphatase 1 (FBPase 1), also known as D-fructose-1, 6-bisphosphate 1-phosphohydrolase 1, and encoded by the gene name FBP1 and FBP, belongs to the FBPase class 1 family. FBPase 1 is a gluconeogenesis regulatory enzyme that catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate in the presence of divalent cations; thus, acting as a rate-limiting enzyme in gluconeogenesis. FBPase 1 has been demonstrated to play a role in regulating glucose sensing and insulin secretion of pancreatic beta-cells. FBPase 1 may be involved in modulating glycerol gluconeogenesis in liver. FBPase 1 has been studied as an important regulator of appetite and adiposity. Increased expression of FBPase 1 in liver, after nutrient excess, increases circulating satiety hormones and reduces appetite-stimulating neuropeptides. Therefore, FBPase 1 may be an important target that provides a feedback mechanism to limit weight gain. FBPase 1 is expressed mainly in pancreatic islets.

Immunogen

KLH-conjugated linear peptide corresponding to human FBPase 1.

Application

Anti-FBPase 1, clone 13A8.1 | MABC543 is an antibody against FBPase 1 for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected FBPase 1 in human pancreas tissue.

Quality

Evaluated by Western Blotting in mouse stomach tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FBPase 1 in 10 µg of mouse stomach tissue lysate.

Target description

~36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia Carolina Leal-Esteban et al.
PLoS genetics, 14(7), e1007487-e1007487 (2018-07-12)
Altered glucose and lipid metabolism fuel cystic growth in polycystic kidneys, but the cause of these perturbations is unclear. Renal cysts also associate with mutations in Bicaudal C1 (Bicc1) or in its self-polymerizing sterile alpha motif (SAM). Here, we found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service