Skip to Content
Merck

MABC1703

Anti-CCL22/MDC Antibody, clone 6F5.7

clone 6F5.7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Clone:
6F5.7, monoclonal
Species reactivity:
-
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


clone

6F5.7, monoclonal

Quality Segment

isotype

IgG2aκ

Immunogen

Recombinant fragment of human CCL22 corresponding to 67 amino acids in the mature protein.

Biochem/physiol Actions

Clone 6F5.7 is a mouse monoclonal antibody that detects human CC-motif chemokine 22 (CCL22).

Preparation Note

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library