Skip to Content
Merck

Skip To

MABC1701

Anti-PDGF-AA Antibody, clone 5C10.8

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
5C10.8, monoclonal
Application:
IHC (p), WB
Citations:
-


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5C10.8, monoclonal

mol wt

calculated mol wt 24.04 kDa, observed mol wt ~28 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

isotype

IgG2bκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDGFA(5154)

General description

Platelet-derived growth factor subunit A (UniProt: P04085d; also known as PDGF subunit A, PDGF-1, Platelet-derived growth factor A chain, Platelet-derived growth factor alpha polypeptide) is encoded by the PDGFA (also known as PDGF1) gene (Gene ID: 5154) in human. PDGFs are a family of four cystine-knot-type growth factors (PDGF-A, -B, -C and -D) that are dimeric mitogenic signaling molecules. In the early developmental stage, they drive the proliferation of undifferentiated mesenchyme and some progenitor populations. During later maturation stages they participate in tissue remodeling, cellular differentiation, and morphogenesis. PDGF A and B can exist as two A subunits (PDGF-AA), two B subunits (PDGF-BB), or one of each (PDGF-AB). These dimeric forms are differentially expressed in various cell types. Their action is mediated via two types of receptors PDGFR and PDGFRβ, which belong to the class III receptor tyrosine kinases and exhibit different expression patterns and physiological roles. PDGFR signaling controls gastrulation and the development of several organs such as lung, intestine, skin, testis, kidney, bones, and neuroprotective tissues and PDGFRβ signaling is an essential regulator of early hematopoiesis and blood vessel formation. PDGF-A is homodimeric protein that is synthesized with a signal peptide (aa 1-20) and a propeptide (aa 21-86) that are subsequently cleaved off to produce the mature protein (aa 87-211). Its signaling activity is modulated by the formation of heterodimers with PDGFB. It plays an essential role in the regulation of embryonic development, cell proliferation, cell migration, survival, and chemotaxis. It is also required for normal lung alveolar septum formation during embryogenesis, normal development of the gastrointestinal tract, normal development of Leydig cells and spermatogenesis. It is also involved in normal oligodendrocyte development and normal myelination in the spinal cord and cerebellum.

Immunogen

Recombinant full-length human Platelet derived growth factor A (without the signal peptide and propeptide).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant human PDGF-AA.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant human PDGF-AA.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected PDGF-AA in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-PDGF-AA, clone 5C10.8, Cat. No. MABC1701, is a mouse monoclonal antibody that detects PDGF-AA and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Biochem/physiol Actions

Clone 5C10.8 is a mouse monoclonal antibody that specifically detects Platelet derived growth factor AA (PDGF-AA).

Physical form

Purified mouse monoclonal antibody IgG2b in PBS with 0.05% sodium azide.

Preparation Note

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters