MABC1207
Anti-CIRP (CIRBP) Antibody, clone 6C7.1
clone 6C7.1, from mouse
Synonym(s):
Cold-inducible RNA-binding protein, A18 hnRNP, Glycine-rich RNA-binding protein CIRP
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6C7.1, monoclonal
species reactivity
rat, human
species reactivity (predicted by homology)
mouse (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... CIRBP(1153)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected CIRP (CIRBP) in 10 µg of Raji human B-cell lymphoma cell lysate.
Apoptosis & Cancer
Quality
Western Blotting Analysis: A 1:2,000 dilution of this antibody detected CIRP (CIRBP) in 10 µg of SH-SY5Y human neuroblastoma cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service