Skip to Content
Merck

Skip To

MABC1193

Anti-LC3A/LC3B Antibody

mouse monoclonal, 4F11.1

Synonym(s):

Microtubule-associated proteins 1A/1B light chain 3B, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, MAP1 light chain 3-like protein 2, MAP1A/MAP1B light chain 3 B, MAP1A/MAP1B LC3 B, Microtubule-associated protein 1 li

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Please contact Customer Service for Availability
4 190,00 CZK

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
4F11.1, monoclonal
Species reactivity:
human
Application:
WB
Citations:
-

4 190,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-LC3A/B Antibody, clone 4F11.1, clone 4F11.1, from mouse

biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F11.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MAP1LC3B(81631)

General description

Microtubule-associated proteins 1A/1B light chain 3B (UniProt: Q9GZQ8; also known as Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, MAP1 light chain 3-like protein 2, MAP1A/MAP1B light chain 3 B, MAP1A/MAP1B LC3 B, Microtubule-associated protein 1 light chain 3 beta) is encoded by the MAP1LC3B (also known as MAP1ALC3) gene (Gene ID: 81631) in human. Autophagy-related protein LC3 A/B is an ubiquitin-like modifier that is involved in formation of autophagosomal vacuoles (autophagosomes). It plays a role in mitophagy that contributes to regulate mitochondrial quantity and quality by eliminating the mitochondria to a basal level to fulfill cellular energy requirements and preventing excess reactive oxygen species production. It contains a microtubule-associated proteins 1A/1B light chain 3B (aa 2-120) and a propeptide region (aa 121-125) that is cleaved off in mature form. The precursor molecule is cleaved by ATG4B (between aa 120-121) to form the cytosolic form LC3-I, which is activated by ATG7 and then transferred to ATG3 and conjugated to phosphatidylethanolamine (PE) to generate the membrane-bound LC3-II. Phosphorylation at threonine12 by PKA inhibits conjugation to phosphatidylethanolamine (PE). Autophagy-related protein LC3 B is found to be most abundant in heart, brain, skeletal muscle and testis.
~15 kDa observed; 14.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST/His-tagged recombinant fragment corresponding to amino acids 2-74 of human Autophagy-related protein LC3 B.

Application

Anti-LC3A/B, clone 4F11.1, Cat. No. MABC1193, is a mouse monoclonal antibody that detects LC3A/B and has been tested for use in Western Blotting.
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

Clone 4F11.1 detects Autophagy-related protein LC3 B in human cells.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cells treated with 50 µM chloroquine.

Western Blotting Analysis: 1 µg/mL of this antibody detected LC3A/B in 10 µg of lysate from HeLa cells treated with 50 µM chloroquine.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L7543L8918ZRB1125
description

clone 4F11.1, from mouse

description

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

description

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

description

recombinant, expressed in HEK 293 cells

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... MAP1LC3B(81631)

Gene Information

human ... MAP1LC3B(81631)
mouse ... Map1lc3b(67443)
rat ... Map1lc3b(64862)

Gene Information

human ... MAP1LC3A(84557), MAP1LC3B(81631)
mouse ... Map1lc3a(66734), Map1lc3b(67443)
rat ... Map1lc3a(362245), Map1lc3b(64862)

Gene Information

-

clone

4F11.1, monoclonal

clone

polyclonal

clone

polyclonal

clone

recombinant monoclonal

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

species reactivity

human

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

technique(s)

western blot: 1-2 μg/mL using whole extracts of rat and mouse brain, indirect immunofluorescence: 5-10 μg/mL using human HeLa cells

technique(s)

western blot: 1-2 μg/mL using whole extracts of rat and mouse brain, immunohistochemistry: suitable, immunoprecipitation (IP): 5-10 μg using extracts of human U87 cells

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunocytochemistry: suitable, western blot: suitable, affinity binding assay: suitable


Still not finding the right product?

Explore all of our products under Anti-LC3A/B Antibody, clone 4F11.1

— or —

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters