MABC1167
Anti-P53DINP1 Antibody, clone F8
from rat
Synonym(s):
TP53INP1, Tumor protein p53-inducible nuclear protein 1, Stress-induced protein, p53-dependent damage-inducible nuclear protein 1
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
monoclonal
species reactivity
human, mouse
packaging
antibody small pack of 25 μL
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG2a
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... TP53INP1(94241)
General description
Specificity
Immunogen
Application
Apoptosis & Cancer
ELISA Analysis: A representative lot detected P53DINP1 in Elisa applications (Saadi, H., et. al. (2013). Results Immunol. 3:51-6).
Western Blotting Analysis: A representative lot detected P53DINP1 in Western Blotting applications (Al Saati, T., et. al. (2013). Am J Pathol. 182(6):1996-2004; Peuget, S., et. al. (2014). Cell Death Differ. 21(7):1107-18; Saadi, H., et. al. (2013). Results Immunol. 3:51-6; Seillier, M., et. al. (2012). Cell Death Differ. 19(9):1525-35).
Quality
Western Blotting Analysis: A 1:62.5 dilution of this antibody detected P53DINP1 in 10 µg of human colon tissue lysate.
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service