Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 μg | Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 4,040.00 |
100 μg | Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 10,500.00 |
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1G6-D7, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
ELISA: suitable, immunoprecipitation (IP): suitable, inhibition assay: suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... HSP90AA1(3320)
General description
Immunogen
Application
ELISA Analysis: A representative lot detected Hsp90 alpha in ELISA applications (Zou, M., et. al. (2017). Oncogene. 36(15):2160-2171).
Affects Function Analysis: A representative lot blocked parental MDA-MB-231 cell invasion in a dose-dependent manner. (Zou, M., et. al. (2017). Oncogene. 36(15):2160-2171).
Western Blotting Analysis: A representative lot detected Hsp90 alpha in Western Blotting applications (Zou, M., et. al. (2017). Oncogene. 36(15):2160-2171).
Inhibits Activity/Function Analysis: A representative lot inhibited both de novo tumor formation and expansion of already formed tumors in mice. (Zou, M., et. al. (2017). Oncogene. 36(15):2160-2171).
Biochem/physiol Actions
Physical form
Analysis Note
Western Blotting Analysis: 1 µg/mL of this antibody detected Hsp90 alpha in MDA-MB-231 cell lysate.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| clone 1G6-D7, monoclonal | clone 2H9, monoclonal | clone polyclonal | clone polyclonal |
| Gene Information human ... HSP90AA1(3320) | Gene Information human ... HSPA1A(3303) | Gene Information human ... HSPD1(3329) | Gene Information human ... HSP90AA1(3320) |
| species reactivity human | species reactivity bovine, human | species reactivity human, rat, mouse | species reactivity human, mouse, rat |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source mouse | biological source mouse | biological source rabbit | biological source rabbit |
| antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



