Skip to Content
Merck

Skip To

MAB4444

Anti-Epithelial Cell Adhesion Molecule (EpCAM) Antibody, clone 8

clone mab 8, from mouse

Synonym(s):

Adenocarcinoma-associated antigen, CD326 antigen, Cell surface glycoprotein Trop-1, Epithelial cell surface antigen, Epithelial glycoprotein, KS 1/4 antigen, Major gastrointestinal tumor-associated protein GA733-2, antigen identified by monoclonal antibo

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 23, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 12,600.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
mab 8, monoclonal
Application:
FACS, ICC, IF, WB
Citations:
-

CZK 12,600.00


Estimated to ship onJune 23, 2026Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1215

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

mab 8, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Epithelial Call Adhesion Molecule (EpCAM, CD326) is a transmembrane glycoprotein of approximately 40 kDa. EpCAM was initially identified as a tumor associated marker having high expression in quickly proliferating epithelial tumors. As such, EpCAM has been of significant interest as a means for tumor diagnosis and therapy. EpCAM is also expressed in normal epithelial cells, but in comparatively lower levels. Preliminary studies limited EpCAM’s function to cell-cell adhesion; however more recent studies indicate it to be involved in diverse processes that include cell migration, cell signaling, differentiation and proliferation.
~ 40 kDa

Immunogen

Epitope: Unknown
HES-3 Cells (Choo, et al. (2008) Stem Cells. Jun;26(6):1454-63)

Application

Flow Cytometry Analysis: 0.5 µg from a previous lot detected Epithelial cell adhesion molecule in human embryonic stem cells.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected EpCAM in human liver tissue, iPSC-derived liver organoid progenitor, and mature liver organoid sections.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected EpCAM in human breast tissue and patient-derived triple negative breast cancer organoid sections.

Immunofluorescence Analysis: A 1:200 dilution from a representative lot detected EpCAM in human small intestine, human colon, duodenum intestinal organoid and colon intestinal organoid tissue sections.

Western Blot Analysis: A previous lot was used by an independent laboratory in WB. (Ng, V., et al., 2010)
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Adhesion (CAMs)
This Anti-Epithelial Cell Adhesion Molecule (EpCAM) Antibody, clone 8 is validated for use in FC, IC, WB for the detection of Epithelial Cell Adhesion Molecule (EpCAM).

Biochem/physiol Actions

This antibody recognizes epithelial cell adhesion molecule (EpCAM), specifically on undifferentiated human embryonic stem cells.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human embryonic stem cells
Evaluated by Immunocytochemistry in human embryonic stem cells.

Immunocytochemistry Analysis: 1-10 µg/mL from a previous lot detected epithelial cell adhesion molecule (EpCAM) in human embryonic stem cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200704SAB4200690ZRB1215
clone

mab 8, monoclonal

clone

Ber-EP4, monoclonal

clone

Ber-EP4, monoclonal

clone

1B8, recombinant monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit (recombinant)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

culture supernatant

antibody form

purified antibody

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters