Skip to Content
Merck
All Photos(1)

Documents

MAB4150

Sigma-Aldrich

Anti-MRP2 Antibody, CT, clone M2 III-6

culture supernatant, clone M2 III-6, Chemicon®

Synonym(s):

cMOAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

M2 III-6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ABCC2(1244)

Specificity

This antibody reacts with an internal epitope of MRP2, a 190-200 kDa transmembrane protein previously known as the canalicular multi-organic anion transporter (cMOAT). MRP2 is a member of the MRP family of multidrug resistance related proteins, and MRP2 over expression has been observed in a subset of cisplatin resistant cell lines. This antibody does not cross-react with the human MDR1, MRP1, MRP3 or MRP5 gene products.

Immunogen

Bacterial fusion protein of MRP2 containing the 202-aa C-terminal end
Epitope: C-terminus

Application

Detect MRP2 using this Anti-MRP2 Antibody, C-terminus, clone M2 III-6 validated for use in WB, IC, IH, IH(P).
Research Category
Metabolism
Research Sub Category
Toxicology & Drug Resistance
Western blotting: 1:20 - 1:50 with anti-mouse HRP

Immunocytochemistry: 1:20 - 1:50 on acetone fixed cytospin preparations

Immunohistochemistry: 1:20 on acetone fixed frozen tissue sections; 1:20 on formaldehyde-fixed paraffin-embedded tissues (pretreat with 0.01M citrate, pH 6.0, in dH2O at 100ºC for 5 minutes, repeated 3 times)

Optimal working dilutions must be determined by end user.

Physical form

Liquid containing 0.7% BSA and 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for up to 6 months. For long-term storage store at -20°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C C Paulusma et al.
Science (New York, N.Y.), 271(5252), 1126-1128 (1996-02-23)
The human Dubin-Johnson syndrome and its animal model, the TR(-) rat, are characterized by a chronic conjugated hyperbilirubinemia. TR(-) rats are defective in the canalicular multispecific organic anion transporter (cMOAT), which mediates hepatobiliary excretion of numerous organic anions. The complementary
Clock and ATF4 transcription system regulates drug resistance in human cancer cell lines.
Igarashi, T; Izumi, H; Uchiumi, T; Nishio, K; Arao, T; Tanabe, M; Uramoto, H; Sugio et al.
Oncogene null
Qinghong Li et al.
Cellular and molecular gastroenterology and hepatology, 14(2), 295-310 (2022-04-15)
UNC45A is a myosin (co-)chaperone, and mutations in the UNC45A gene were recently identified in osteo-oto-hepato-enteric (O2HE) syndrome patients presenting with congenital diarrhea and intrahepatic cholestasis. Congenital diarrhea and intrahepatic cholestasis are also the prime symptoms in patients with microvillus
Cifford W Mason et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(6), 1000-1007 (2011-03-25)
Fetal drug exposure is determined by the type and concentration of placental transporters, and their regulation is central to the development of new treatments and delivery strategies for pregnant women and their fetuses. We tested the expression of several clinically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service